Skip to Content
Merck
All Photos(1)

Key Documents

D1065

Sigma-Aldrich

4-(2-[6-(Dioctylamino)-2-naphthalenyl]ethenyl)-1-(3-sulfopropyl)pyridinium inner salt

≥95% purity (HPLC), solid

Synonym(s):

Di-8-ANEPPS

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H52N2O3S
CAS Number:
Molecular Weight:
592.87
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Product Name

4-(2-[6-(Dioctylamino)-2-naphthalenyl]ethenyl)-1-(3-sulfopropyl)pyridinium inner salt, ≥95% (HPLC), solid

Quality Level

Assay

≥95% (HPLC)

form

solid

mol wt

592.88

color

orange to red

solubility

DMSO: soluble
ethanol: soluble

ε (extinction coefficient)

≥37 at 495-501  nm in methanol

fluorescence

λex 710-716 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-8°C

SMILES string

CCCCCCCCN(CCCCCCCC)c1ccc2cc(\C=C\c3cc[n+](CCCS([O-])(=O)=O)cc3)ccc2c1

InChI

1S/C36H52N2O3S/c1-3-5-7-9-11-13-25-38(26-14-12-10-8-6-4-2)36-21-20-34-30-33(18-19-35(34)31-36)17-16-32-22-27-37(28-23-32)24-15-29-42(39,40)41/h16-23,27-28,30-31H,3-15,24-26,29H2,1-2H3

InChI key

IXFSUSNUALIXLU-UHFFFAOYSA-N

Biochem/physiol Actions

Fast-responding membrane potential dye.

Linkage

More suitable for long-term membrane potential studies than di-4-ANEPPS (Cat. No. D8064) because of better retention in the outer leaflet of plasma membranes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sanaya N Shroff et al.
iScience, 23(4), 100974-100974 (2020-04-17)
Precise measurement of action potentials (APs) is needed to observe electrical activity and cellular communication within cardiac tissue. Voltage-sensitive dyes (VSDs) are traditionally used to measure cardiac APs; however, they require acute chemical addition that prevents chronic imaging. Genetically encoded
Antonio Celestino-Montes et al.
Life sciences, 264, 118700-118700 (2020-11-02)
Ventricular myocytes (VM) depolarization activates L-type Ca2+ channels (LCC) allowing Ca2+ influx (ICa) to synchronize sarcoplasmic reticulum (SR) Ca2+ release, via Ca2+-release channels (RyR2). The resulting whole-cell Ca2+ transient triggers contraction, while cytosolic Ca2+ removal by SR Ca2+ pump (SERCA2)
Kirby R Campbell et al.
The journal of physical chemistry. B, 121(8), 1749-1757 (2017-02-09)
Extensive remodeling of the extracellular matrix (ECM) occurs in many epithelial cancers. For example, in ovarian cancer, upregulation of collagen isoform type III has been linked to invasive forms of the disease, and this change may be a potential biomarker.
Kate Hsu et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 31(10), 4256-4264 (2017-06-10)
Human CO
Carlos A Valverde et al.
Cardiovascular research, 115(3), 556-569 (2018-09-01)
Abnormal Ca2+ release from the sarcoplasmic reticulum (SR), associated with Ca2+-calmodulin kinase II (CaMKII)-dependent phosphorylation of RyR2 at Ser2814, has consistently been linked to arrhythmogenesis and ischaemia/reperfusion (I/R)-induced cell death. In contrast, the role played by SR Ca2+ uptake under

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service