Skip to Content
Merck
All Photos(2)

Documents

B12695

Sigma-Aldrich

Benzoyl chloride

ReagentPlus®, ≥99%

Synonym(s):

Benzoic acid chloride, alpha-chlorobenzaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COCl
CAS Number:
Molecular Weight:
140.57
Beilstein:
471389
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

vapor density

4.88 (vs air)

Quality Level

vapor pressure

1 mmHg ( 32 °C)

product line

ReagentPlus®

Assay

≥99%

form

liquid

autoignition temp.

1056 °F

expl. lim.

4.9 %

refractive index

n20/D 1.553 (lit.)

bp

198 °C (lit.)

mp

−1 °C (lit.)

density

1.211 g/mL at 25 °C (lit.)

SMILES string

ClC(=O)c1ccccc1

InChI

1S/C7H5ClO/c8-7(9)6-4-2-1-3-5-6/h1-5H

InChI key

PASDCCFISLVPSO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Benzoyl chloride, an acid chloride, is a commonly used benzoylating agent. The arylation of 1,1- and 1,2-disubstituted activated alkenes using benzoyl chloride in the presence of palladium catalyst has been investigated.

Application

Benzoyl chloride may be used to synthesize:
  • 4-Methoxybenzophenone via acylation of anisole in the presence of cesium-substituted dodecatungstophosphoric acid (Cs2.5H0.5PW12O40) supported on K-10 clay.
  • Benzoylamino acids via acylation of amino acids in the presence of diisopropylethylamine.
  • p-, o-, m-Methylbenzophenones via benzoylation of toluene in the presence of Al-promoted sulfated zirconia catalysts.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

161.6 °F - closed cup

Flash Point(C)

72 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benzoylation of toluene with benzoyl chloride on Al-promoted sulfated solid superacids.
Xia Y, et al.
Catalysis Letters, 55(2), 105-112 (1998)
Benzoylation of anisole over borate zirconia solid acid catalyst.
Patil PT, et al.
Catalysis Communications, 3(9), 411-416 (2002)
Cesium-substituted dodecatungstophosphoric acid on K-10 clay for benzoylation of anisole with benzoyl chloride.
Yadav GD, et al.
J. Catal., 217(1), 88-99 (2003)
Stereochemical course of the palladium-catalysed arylation of disubstituted activated alkenes with benzoyl chloride.
Spencer A.
Journal of Organometallic Chemistry, 240(2), 209-216 (1982)
Nils C Gassen et al.
Nature communications, 12(1), 3818-3818 (2021-06-23)
Viruses manipulate cellular metabolism and macromolecule recycling processes like autophagy. Dysregulated metabolism might lead to excessive inflammatory and autoimmune responses as observed in severe and long COVID-19 patients. Here we show that SARS-CoV-2 modulates cellular metabolism and reduces autophagy. Accordingly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service