Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Benzoyl chloride
Synonym(s):
Benzoyl chloride
Linear Formula:
C
6
H
5
COCl
CAS Number:
98-88-4
Molecular Weight:
140.57
EC Number:
202-710-8
Products
Product Category
building blocks (1)
Brand
Sigma-Aldrich (4)
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Markush Class
Markush Group
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
259950
Benzoyl chloride,
99%
Expand
View Pricing
8.01804
Benzoyl chloride,
for synthesis
Expand
View Pricing
B12695
Benzoyl chloride,
ReagentPlus
®
, ≥99%
Expand
View Pricing
03-1440
Benzoyl chloride,
SAJ first grade, ≥98.0%
Expand
View Pricing