Skip to Content
Merck
All Photos(1)

Documents

B1054000

Betamethasone 17-valerate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

1,4-Pregnadiene-11β,17α,21-triol-9α-fluoro-16β-methyl-3,20-dione 17-valerate, 9α-Fluoro-16β-methyl-11β,17α,21-trihydroxy-1,4-pregnadiene-3,20-dione 17-valerate, 9α-Fluoro-16β-methylprednisolone 17-valerate, Betnovate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H37FO6
CAS Number:
Molecular Weight:
476.58
Beilstein:
4240001
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

betamethasone

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CCCCC(=O)O[C@@]1([C@@H](C)CC2C3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C)C(=O)CO

InChI

1S/C27H37FO6/c1-5-6-7-23(33)34-27(22(32)15-29)16(2)12-20-19-9-8-17-13-18(30)10-11-24(17,3)26(19,28)21(31)14-25(20,27)4/h10-11,13,16,19-21,29,31H,5-9,12,14-15H2,1-4H3/t16-,19?,20?,21-,24-,25-,26-,27-/m0/s1

InChI key

SNHRLVCMMWUAJD-QDHNOTTGSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Betamethasone 17-valerate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Feray Soyupek et al.
Rheumatology international, 32(10), 3229-3234 (2011-11-01)
Firstly, we aimed to determine the effectiveness of various treatment modalities using ultrasonography (US), and secondly, we aimed to assess the correlations between the ultrasonographic findings and electrophysiological tests, symptom severity, functional status and physical findings. 74 hands of 47
V Salini et al.
International journal of immunopathology and pharmacology, 24(1), 211-216 (2011-04-19)
Relapses are frequently observed in subjects with chronic tendinopathies. Corticosteroid injections are usually performed with positive results, but are uncomfortable for the patient and not free from side effects. The aim of this pilot study is to evaluate the short-term
Jin Zhang et al.
Journal of pharmaceutical sciences, 100(3), 896-903 (2010-09-15)
Corticosteroids are therapeutic agents widely used in the pharmacological treatment of skin diseases such as eczema or psoriasis. Unfortunately, their use is restricted by the side effects that frequently occur at the systemic level. The goal of the research described
Luigi Naldi et al.
American journal of clinical dermatology, 12(3), 191-201 (2011-02-03)
Corticosteroids are a versatile option for the treatment of mild-to-moderate psoriasis due to their availability in a wide range of potencies and formulations. Occlusion of the corticosteroid is a widely accepted procedure to enhance the penetration of the medication, thereby
Annabel Maruani et al.
International journal of pharmaceutics, 395(1-2), 84-90 (2010-05-18)
Topical steroids are efficient in vasoconstriction potential, which is linked to their anti-inflammatory activity. Low-frequency ultrasound (US) applied on the skin (sonophoresis) may enhance the transdermal transport of various steroids. We aimed to assess, in a simple, blinded, randomized controlled

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service