Skip to Content
Merck
All Photos(1)

Key Documents

B1040000

Betamethasone dipropionate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Betamethasone 17,21-dipropionate, 9α-Fluoro-16β-methyl-11β,17α,21-trihydroxy-1,4-pregnadiene-3,20-dione 17,21-dipropionate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H37FO7
CAS Number:
Molecular Weight:
504.59
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

betamethasone

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[H][C@@]12C[C@H](C)[C@](OC(=O)CCl)(C(=O)COC(=O)CC)[C@@]1(C)C[C@H](O)C3(F)[C@@]2([H])CCC4=CC(=O)C=C[C@]34C

InChI

1S/C27H34ClFO7/c1-5-22(33)35-14-21(32)27(36-23(34)13-28)15(2)10-19-18-7-6-16-11-17(30)8-9-24(16,3)26(18,29)20(31)12-25(19,27)4/h8-9,11,15,18-20,31H,5-7,10,12-14H2,1-4H3/t15-,18-,19-,20-,24-,25-,26?,27-/m0/s1

InChI key

UOCNBCVPZWLUKX-HOBFILNXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the Issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Betamethasone dipropionate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Betamethasone 17,21-dipropionate is a glucocorticoid with anti-inflammatory and immunosuppressive activity.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B - STOT RE 2

Target Organs

Liver,Kidney,Endocrine system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Catherine Queille-Roussel et al.
The Journal of dermatological treatment, 24(4), 250-254 (2011-11-23)
The efficacy and safety of calcipotriol plus betamethasone dipropionate ointment in the treatment of psoriasis vulgaris has consistently been demonstrated in several clinical trials. For treatment of scalp psoriasis, more convenient formulations are required. Therefore, new lipophilic alcohol-free gel formulations
Gino A Vena et al.
The Journal of dermatological treatment, 23(4), 255-260 (2011-07-16)
Combination therapy is a common approach to psoriasis, aimed at improving clinical response and minimizing the risk of side effects. The aim of this pilot randomized open-label study was to evaluate the efficacy and safety of the combination of low-dose
Mohd Hanif Zulfakar et al.
International journal of pharmaceutics, 434(1-2), 399-405 (2012-06-14)
The current work examined the effect of fish oil (FO) and betamethasone dipropionate (BD) on the growth of immortalized HaCaT keratinocytes. HaCaT cells were grown and treated with FO and/or BD, and proliferation determined using the MTT method. The cells
Naresh Punwani et al.
Journal of the American Academy of Dermatology, 67(4), 658-664 (2012-01-28)
Janus-associated kinases (JAKs) are involved in signal transduction from a variety of cytokines implicated in the pathogenesis of psoriasis, including interleukin (IL)-12, IL-23, and interferon-γ. INCB018424, a small molecule inhibitor of JAK1 and JAK2, inhibits cytokine-induced JAK/signal transducers and activators
Srinath Reddy et al.
Acta paediatrica (Oslo, Norway : 1992), 101(3), e130-e133 (2011-11-23)
Phimosis is a common paediatric urological disorder and often necessitates circumcision. We prospectively evaluated local steroid therapy (LST) as the first choice therapy for such children. Two hundred and sixty symptomatic boys up to 15 years of age (mean 34

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service