Skip to Content
Merck
All Photos(3)

Key Documents

A36883

Sigma-Aldrich

Aurintricarboxylic acid ammonium salt

ACS reagent, powder or crystals

Synonym(s):

ATA, Aluminon, Ammonium aurintricarboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C22H14O9 · 3NH3
CAS Number:
Molecular Weight:
473.43
Colour Index Number:
43810
Beilstein:
3900820
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Product Name

Aurintricarboxylic acid ammonium salt, ACS reagent

biological source

synthetic

Quality Level

grade

ACS reagent

form

powder or crystals

technique(s)

titration: suitable

impurities

≤0.1% insolubles

ign. residue

≤0.2%

color

dark red

mp

220-225 °C (dec.) (lit.)

solubility

water: soluble 100 mg/mL

λmax

450 nm
522 nm (2nd)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

N.N.N.OC(=O)c1cc(ccc1O)\C(c2ccc(O)c(c2)C(O)=O)=C3/C=CC(=O)C(=C3)C(O)=O

InChI

1S/C22H14O9.3H3N/c23-16-4-1-10(7-13(16)20(26)27)19(11-2-5-17(24)14(8-11)21(28)29)12-3-6-18(25)15(9-12)22(30)31;;;/h1-9,23-24H,(H,26,27)(H,28,29)(H,30,31);3*1H3

InChI key

AIPNSHNRCQOTRI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reagent for aluminum

Biochem/physiol Actions

Aurintricarboxylic acid readily polymerizes in aqueous solution, forming a stable free radical that inhibits protein-nucleic acid interactions. It is a potent inhibitor of ribonuclease and topoisomerase II by preventing the binding of the nucleic acid to the enzyme. It stimulates tyrosine phosphorylation processes including the Jak2/STAT5 pathway in NB2 lymphoma cells, ErbB4 in neuroblastoma cells, and MAP kinases, Shc proteins, phosphatidylinositide 3-kinase and phospholipase C in PC12 cells. Aurintricarboxylic acid inhibits apoptosis in many cell types. Its neuroprotective effect, perhaps due to its ability to prevent down-regulation of Ca2+ impermeable GluR2 receptors or to its ability to inhibit calpain, a Ca2+-activated protease that is activated during apoptosis.

Other Notes

May contain a substantial amount of polymeric material.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hui-Chen Hung et al.
Antiviral research, 81(2), 123-131 (2008-11-19)
There is a continuing threat that the highly pathogenic avian influenza virus will cause future influenza pandemics. In this study, we screened a library of compounds that are biologically active and structurally diverse for inhibitory activity against influenza neuraminidase (NA).
Jakyung Yoo et al.
Advances in protein chemistry and structural biology, 87, 219-247 (2012-05-23)
DNA methyltransferases (DNMTs) are promising epigenetic targets for the development of novel anticancer drugs and other diseases. Molecular modeling and experimental approaches are being used to identify and develop inhibitors of human DNMTs. Most of the computational efforts conducted so
Hui-Chen Hung et al.
The Journal of antimicrobial chemotherapy, 65(4), 676-683 (2010-01-22)
Enterovirus 71 (EV71) causes serious diseases in humans. The aim of this study was to examine the effects of aurintricarboxylic acid (ATA) on EV71 replication and to explore the underlying mechanism. To measure the activity of ATA in inhibiting the
Mélanie K Rich et al.
Plant physiology, 168(3), 788-797 (2015-05-15)
Arbuscular mycorrhiza (AM) is a mutual symbiosis that involves a complex symbiotic interface over which nutrients are exchanged between the plant host and the AM fungus. Dozens of genes in the host are required for the establishment and functioning of
Qiang Peng et al.
PloS one, 7(10), e47273-e47273 (2012-11-03)
Baboons receiving xenogeneic livers from wild type and transgenic pigs survive less than 10 days. One of the major issues is the early development of profound thrombocytopenia that results in fatal hemorrhage. Histological examination of xenotransplanted livers has shown baboon

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service