Skip to Content
Merck
All Photos(1)

Key Documents

94145

Supelco

7α-Hydroxy-4-cholesten-3-one-25,26,26,26,27,27,27-d7

≥95.0% (HPLC)

Synonym(s):

7α-Hydroxycholest-4-en-3-one-d7

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27D7H37O2
Molecular Weight:
407.68
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

application(s)

clinical testing

format

neat

storage temp.

−20°C

SMILES string

O=C1CC[C@@]2(C)C(C[C@@H](O)[C@]3([H])[C@]2([H])CC[C@@]4(C)[C@@]3([H])CC[C@]4([H])[C@H](C)CCCC(C([2H])([2H])[2H])([2H])C([2H])([2H])[2H])=C1

InChI key

IOIZWEJGGCZDOL-CTNVFXSSSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W Gordon Brydon et al.
Canadian journal of gastroenterology = Journal canadien de gastroenterologie, 25(6), 319-323 (2011-07-19)
Bile acid malabsorption (BAM) is a recognized cause of watery diarrhea, often diagnosed empirically based on clinical response to cholestyramine. The radionuclide selenium-labelled homocholic acid-taurine whole body retention test is expensive, labour intensive and of limited availability. To report on
Chronic diarrhea due to excessive bile acid synthesis and not defective ileal transport: a new syndrome of defective fibroblast growth factor 19 release.
Alan F Hofmann et al.
Clinical gastroenterology and hepatology : the official clinical practice journal of the American Gastroenterological Association, 7(11), 1151-1154 (2009-08-12)
Akira Honda et al.
Journal of lipid research, 48(2), 458-464 (2006-11-10)
We describe a highly sensitive and specific method for the quantification of serum 7alpha-hydroxy-4-cholesten-3-one (C4), which has been used as a biomarker for bile acid biosynthesis. This method is based upon a stable isotope dilution technique by liquid chromatography-tandem mass
Cecilia Gälman et al.
Gastroenterology, 129(5), 1445-1453 (2005-11-16)
The conversion of cholesterol to bile acids by the liver is an important regulator of body cholesterol homeostasis. In rodents, both cholesterol and bile acid synthesis have marked diurnal rhythms that peak synchronously at midnight. The aim of this study
David W Russell
Annual review of biochemistry, 72, 137-174 (2003-01-25)
The synthesis and excretion of bile acids comprise the major pathway of cholesterol catabolism in mammals. Synthesis provides a direct means of converting cholesterol, which is both hydrophobic and insoluble, into a water-soluble and readily excreted molecule, the bile acid.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service