Skip to Content
Merck
All Photos(1)

Key Documents

90268

Sigma-Aldrich

Polyvinylpyrrolidone

K 25, tested according to Ph. Eur.

Synonym(s):

PVP, Polyvidone, Povidone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H9NO)n
CAS Number:
MDL number:
UNSPSC Code:
12352005
NACRES:
NA.21

Agency

tested according to Ph. Eur.

form

powder

SMILES string

C=CN1CCCC1=O

InChI

1S/C6H9NO/c1-2-7-5-3-4-6(7)8/h2H,1,3-5H2

InChI key

WHNWPMSKXPGLAX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Colyophilized Sugar-Polymer Dispersions for Enhanced Processing and Storage Stability.: This study explores the use of polyvinylpyrrolidone in colyophilized sugar-polymer dispersions to improve the processing and storage stability of pharmaceuticals. The research demonstrates how combining sugars with polyvinylpyrrolidone enhances the physical stability and performance of the resulting dispersions, which is crucial for maintaining drug efficacy during storage and handling (Giannachi et al., 2024).

  • Fabrication and evaluation of stable amorphous polymer-drug composite particles via a nozzle-free ultrasonic nebulizer.: This article discusses the development of amorphous polymer-drug composite particles using polyvinylpyrrolidone and a nozzle-free ultrasonic nebulizer. The findings indicate that polyvinylpyrrolidone helps in stabilizing the amorphous form of the drug, which can enhance its solubility and bioavailability (Lee et al., 2024).

  • Polymer-Assisted Coprecipitation Synthesized Zinc Oxide Nanoparticles and Their Uses for Gre en Chemical Synthesis via Photocatalytic Glucose Conversions.: This study presents a novel method of using polyvinylpyrrolidone in the synthesis of zinc oxide nanoparticles. The research shows that these nanoparticles can be used in green chemical processes, such as the photocatalytic conversion of glucose, showcasing the versatility of polyvinylpyrrolidone in catalysis and material science (Kaewsaenee et al., 2023).

  • Development and evaluation of the in vitro schistosomicidal activity of solid dispersions based on 2-(-5-bromo-1-H-indole-3-yl-methylene)-N-(naphthalene-1-ylhydrazine-carbothiamide).: This article explores the use of polyvinylpyrrolidone in formulating solid dispersions for antiparasitic treatments. The findings demonstrate that polyvinylpyrrolidone can enhance the solubility and efficacy of the active pharmaceutical ingredient against parasitic infections (de Luna Rocha et al., 2024).

Other Notes

Polyvinylpyrrolidone is a component of Denhardt′s Solution and is included at a concentration of 1% (w/v) in the standard 50X stock solution.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jennifer Quirós et al.
Journal of hazardous materials, 299, 298-305 (2015-07-05)
The use of electrospun polyvinylpyrrolidone (PVP) nanofibers containing silver, copper, and zinc nanoparticles was studied to prepare antimicrobial mats using silver and copper nitrates and zinc acetate as precursors. Silver became reduced during electrospinning and formed nanoparticles of several tens
Na0.44 MnO2 with very fast sodium diffusion and stable cycling synthesized via polyvinylpyrrolidone-combustion method.
Dai K, et al.
Journal of Power Sources, 285, 161-168 (2015)
Kallum M Koczkur et al.
Dalton transactions (Cambridge, England : 2003), 44(41), 17883-17905 (2015-10-06)
Colloidal synthesis offers a route to nanoparticles (NPs) with controlled composition and structural features. This Perspective describes the use of polyvinylpyrrolidone (PVP) to obtain such nanostructures. PVP can serve as a surface stabilizer, growth modifier, nanoparticle dispersant, and reducing agent.
Markéta Julinová et al.
Water environment research : a research publication of the Water Environment Federation, 84(12), 2123-2132 (2013-01-25)
Polyvinylpyrrolidone (PVP) is a frequently used polymer in the pharmaceutical and foodstuff industries. Because it is not subject to metabolic changes and is virtually nondegradable, trace concentrations of PVP are often found in community wastewaters. The literature finds that the
K Yamashita et al.
Die Pharmazie, 68(1), 54-57 (2013-03-01)
The fullerene C60 is used in consumer products such as cosmetics owing to its antioxidative effects and is being developed for nanomedical applications. However, knowledge regarding the safety of fullerene C60, especially after oral administration, is sparse. Here, we examined

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service