Skip to Content
Merck
All Photos(1)

Key Documents

77500

Supelco

1,10-Phenanthroline monohydrate

for the spectrophotometric determination of Fe, Pd, V, ≥99.0%

Synonym(s):

o-Phenanthroline monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H8N2 · H2O
CAS Number:
Molecular Weight:
198.22
Beilstein:
4008096
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NA.21
Pricing and availability is not currently available.

Quality Level

Assay

≥99.0% (HPLC)
≥99.0% (NT)
≥99.0%

form

crystals

quality

for the spectrophotometric determination of Fe, Pd, V

technique(s)

UV/Vis spectroscopy: suitable
titration: suitable

ign. residue

≤0.05%

mp

100-104 °C (lit.)
99-104 °C

cation traces

Ca: ≤50 mg/kg
Cd: ≤20 mg/kg
Co: ≤20 mg/kg
Cr: ≤20 mg/kg
Cu: ≤20 mg/kg
Fe: ≤20 mg/kg
K: ≤100 mg/kg
Mg: ≤20 mg/kg
Mn: ≤20 mg/kg
Na: ≤100 mg/kg
Ni: ≤20 mg/kg
Pb: ≤20 mg/kg
Zn: ≤20 mg/kg

SMILES string

O.c1cnc2c(c1)ccc3cccnc23

InChI

1S/C12H8N2.H2O/c1-3-9-5-6-10-4-2-8-14-12(10)11(9)13-7-1;/h1-8H;1H2

InChI key

PPQJCISYYXZCAE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,10-Phenanthroline monohydrate is a chelating agent.[1]

Application

It has been used in preparing 0.1 % phenanthroline solution.[2] In another study, 80μl of 1,10-Phenanthroline monohydrate was used along with nanoparticles to measure iron content using colorimetric assay.3 It may be used in the preparation of [Mn(1,10-phen)2(mal)].2H2O.[3]
When complexed with copper, it possesses nuclease activity that has been used to study DNA-protein interactions.

Biochem/physiol Actions

Metalloprotease inhibitor; chelates iron, zinc and other divalent metals. Effective concentration 1-10 mM.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Generation of magnetic nonviral gene transfer agents and magnetofection in vitro.
Mykhaylyk O
Nature Protocols, 2(10), 2391-2411 (2007)
Olivia L Lanier et al.
International journal of hyperthermia : the official journal of European Society for Hyperthermic Oncology, North American Hyperthermia Group, 36(1), 687-701 (2019-07-26)
Background: Magnetic nanoparticles (MNPs) generate heat when exposed to an alternating magnetic field. Consequently, MNPs are used for magnetic fluid hyperthermia (MFH) for cancer treatment, and have been shown to increase the efficacy of chemotherapy and/or radiation treatment in clinical
Insights into the mode of action of the anti-Candida activity of 1,10-phenanthroline and its metal chelates.
McCann M
Metal-based drugs, 7(4), 185-193 (2000)
Microchemical Journal, Devoted to the Application of Microtechniques in All Branches of Science, 15, 585-585 (1970)
W. Sliwa
Heterocycles, 12, 1207-1207 (1979)

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service