Skip to Content
Merck
All Photos(1)

Key Documents

77299

Supelco

Heptacosafluorotributylamine

analytical standard, suitable for mass spectrometry (MS)

Synonym(s):

PFTBA, Perfluorotributylamine, Tris(nonafluorobutyl)amine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
N(CF2CF2CF2CF3)3
CAS Number:
Molecular Weight:
671.09
Beilstein:
1813883
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

vapor density

23.3 (vs air)

vapor pressure

1.3 mmHg ( 25 °C)

technique(s)

mass spectrometry (MS): suitable

bp

178 °C (lit.)

density

1.883 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

FC(F)(F)C(F)(F)C(F)(F)C(F)(F)N(C(F)(F)C(F)(F)C(F)(F)C(F)(F)F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C12F27N/c13-1(14,7(25,26)27)4(19,20)10(34,35)40(11(36,37)5(21,22)2(15,16)8(28,29)30)12(38,39)6(23,24)3(17,18)9(31,32)33

InChI key

RVZRBWKZFJCCIB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Heptacosafluorotributylamine has been used as a reference standard in the calibration of mass scale of the Q exactive gas chromatography-mass spectrometer used in the analysis of molecular isotopic structures of organics.
Heptacosafluorotributylamine may be used as a calibration standard for the gas chromatography coupled to mass spectrometric determination of the composition and characteristics of short-, medium- and long-chain polychlorinated n-alkane (PCA) mixtures in dust samples.
Heptacosafluorotributylamine has been used as a reference standard in the calibration of mass scale of the Q exactive gas chromatography-mass spectrometer used in the analysis of molecular isotopic structures of organics.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characterization of polychlorinated n-alkanes using comprehensive two-dimensional gas chromatography-electron-capture negative ionisation time-of-flight mass spectrometry
Korytar.P, et al.
Journal of Chromatography A, 1086(1-2), 71-82 (2005)
Analysis of molecular isotopic structures at high precision and accuracy by Orbitrap mass spectrometry
Eiler J, et al.
International Journal of Mass Spectrometry (2017)
Daniela F Duarte Campos et al.
Biofabrication, 5(1), 015003-015003 (2012-11-23)
Over the last decade, bioprinting technologies have begun providing important tissue engineering strategies for regenerative medicine and organ transplantation. The major drawback of past approaches has been poor or inadequate material-printing device and substrate combinations, as well as the relatively
Stanislav S Rubakhin et al.
Analytical chemistry, 75(20), 5374-5380 (2004-01-09)
MALDI MS imaging and single-cell profiling are important new capabilities for mass spectrometry. The distribution of neuropeptides within a cell plays an important role in the functioning of the cells in a neuronal network. Protocols for subcellular MALDI MS are
Nadav Kimelman-Bleich et al.
Biomaterials, 30(27), 4639-4648 (2009-06-23)
A major hurdle to surmount in bone-tissue engineering is ensuring a sufficient oxygen supply to newly forming tissue to avoid cell death or delayed development of osteogenic features. We hypothesized that an oxygen-enriched hydrogel scaffold would enhance tissue-engineered bone formation

Articles

MALDI-TOF MS is a tool for confirming end group identity & transformations, verifying successful & complete conjugation. Functionalized polymers enable a quick & efficient route for synthesis of various polymer conjugates for many applications if apt end group is present.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service