Skip to Content
Merck
All Photos(1)

Key Documents

85440

Sigma-Aldrich

(−)-Sinigrin hydrate

≥99.0% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16KNO9S2 · xH2O
CAS Number:
Molecular Weight:
397.46 (anhydrous basis)
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

plant (Brassica nigra)

Assay

≥99.0% (TLC)

form

powder or crystals

optical activity

[α]20/D −17±1°, c = 1% in H2O

technique(s)

thin layer chromatography (TLC): suitable

color

white to faint beige

mp

128 (dec.) (lit.)

cation traces

K: 8.4-10.8

storage temp.

room temp

SMILES string

[K+].[H]O[H].OC[C@H]1O[C@@H](S\C(CC=C)=N\OS([O-])(=O)=O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C10H17NO9S2.K.H2O/c1-2-3-6(11-20-22(16,17)18)21-10-9(15)8(14)7(13)5(4-12)19-10;;/h2,5,7-10,12-15H,1,3-4H2,(H,16,17,18);;1H2/q;+1;/p-1/b11-6+;;/t5-,7-,8+,9-,10+;;/m1../s1

InChI key

IUBVMJHASFBYGW-WBMBWNLZSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Sinigrin, a glucosinolate, is used as a systrate to identify, differentiate and characterize myrosinase(s)/thioglucoside glucohydrolase(s)/thioglucosidase(s). Sinigrin is used as a reference material in procedures for the isolation and identification of glucosinolates.

Biochem/physiol Actions

A β-D-thioglucopyranoside occurring in black mustard seeds and horseradish root. Substrate for thioglucosidase.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A.J. MacLeod et al.
Phytochemistry, 25, 1047-1047 (1986)
Cong Cong et al.
International journal of molecular medicine, 48(2) (2021-07-20)
The present study investigated the function of sinigrin in angiotensin II (Ang II)‑induced renal damage. The results demonstrated that systolic blood pressure (SBP) and diastolic blood pressure (DBP) were increased in Ang II‑challenged rats, and sinigrin treatment inhibited their increase. The levels of blood
Monika Okulicz
Plant foods for human nutrition (Dordrecht, Netherlands), 65(3), 217-224 (2010-09-03)
Sinigrin (SIN) and allyl isothiocyanate (AITC) are compounds found in high concentrations in Brassica family vegetables, especially in Brussels sprouts. Recently, they have been used as a nutrition supplement for their preventive and medicinal effect on some types of cancer
Richard A Lankau
Proceedings of the National Academy of Sciences of the United States of America, 109(28), 11240-11245 (2012-06-27)
Although reciprocal evolutionary responses between interacting species are a driving force behind the diversity of life, pairwise coevolution between plant competitors has received less attention than other species interactions and has been considered relatively less important in explaining ecological patterns.
Richard A Lankau et al.
Science (New York, N.Y.), 317(5844), 1561-1563 (2007-09-18)
The forces that maintain genetic diversity among individuals and diversity among species are usually studied separately. Nevertheless, diversity at one of these levels may depend on the diversity at the other. We have combined observations of natural populations, quantitative genetics

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service