Skip to Content
Merck
All Photos(1)

Documents

72954

Supelco

1-Heptanol

analytical standard, ≥99.5% (GC)

Synonym(s):

Heptyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)6OH
CAS Number:
Molecular Weight:
116.20
Beilstein:
1731686
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor pressure

0.5 mmHg ( 20 °C)

Assay

≥99.5% (GC)

autoignition temp.

662 °F

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.424 (lit.)
n20/D 1.424

bp

176 °C (lit.)

mp

−36 °C (lit.)

density

0.822 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CCCCCCCO

InChI

1S/C7H16O/c1-2-3-4-5-6-7-8/h8H,2-7H2,1H3

InChI key

BBMCTIGTTCKYKF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-Heptanol may be used as a model compound for the determination of alcohols with fluorescence derivatization reagent, 4-(N,N-dimethylaminosulfonyl)-7-(2-chloroformylpyrrolidin-1-yl)-2,1,3-benzoxadiazole using liquid chromatography technique.
1-Heptanol may be used as internal standard in the determination of 4-ethylphenol and 4-ethylguaiacol, which are volatile phenols in red wine samples using gas chromatography coupled with mass spectrometry(GC-MS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

168.8 °F - closed cup

Flash Point(C)

76 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of alcohols and amines, labelled with 4-(N, N-dimethylaminosulfonyl)-7-(2-chloroformylpyrrolidin-1-yl)-2, 3, 1-benzoxadiazole, by liquid chromatography with conventional and laser-induced fluorescence detection
Toyooka T, et al.
Analytica Chimica Acta, 285(3), 343-351 (1994)
Christine Mayr Marangon et al.
Food chemistry, 344, 128658-128658 (2020-12-07)
The Italian grape pomace distillate grappa is often refined by ageing in wooden barrels. Chemical changes of the volatile profile of two samples produced from a Cabernet Sauvignon/Merlot blend and Prosecco pomace both at two ethanol contents (55% and 68%)
Determination of volatile phenols in red wines by dispersive liquid?liquid microextraction and gas chromatography?mass spectrometry detection
Farina L, et al.
Journal of Chromatography A, 1157, 46-50 (2007)
Bertrand Caillier et al.
Fundamental & clinical pharmacology, 26(5), 599-608 (2011-06-01)
QRS widening and QT prolongation are associated with bupropion. The objectives were to elucidate its cardiac electrophysiological properties. Patch-clamp technique was used to assess the I(Kr) -, I(Ks) -, and I(Na) -blocking effects of bupropion. Langendorff retroperfusion technique on isolated
Brian Bao et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 25(1), 255-264 (2010-09-30)
Microtissue self-assembly is thought to be driven primarily by cadherins, while connexons have been examined mainly in intercellular coupling. We investigated whether connexon 43 (Cx43)-mediated cell adhesion modulates self-assembly of human KGN granulosa cells, normal human fibroblasts (NHFs), and MCF-7

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service