Skip to Content
Merck
All Photos(1)

Documents

293075

Sigma-Aldrich

Succinic acid-2,2,3,3-d4

98 atom % D

Synonym(s):

Butanedioic acid-2,2,3,3-d4

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CCD2CD2CO2H
CAS Number:
Molecular Weight:
122.11
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % D

Quality Level

Assay

99% (CP)

form

solid

bp

235 °C

mp

187-190 °C (lit.)

mass shift

M+4

SMILES string

[2H]C([2H])(C(O)=O)C([2H])([2H])C(O)=O

InChI

1S/C4H6O4/c5-3(6)1-2-4(7)8/h1-2H2,(H,5,6)(H,7,8)/i1D2,2D2

InChI key

KDYFGRWQOYBRFD-LNLMKGTHSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marta Consegal et al.
International journal of molecular sciences, 22(8) (2021-05-01)
Succinate dehydrogenase (SDH) inhibition with malonate during reperfusion reduced myocardial infarction in animals, whereas its endogenous substrate, succinate, is detected in plasma from STEMI patients. We investigated whether protection by SDH inhibition is additive to that of remote ischemic perconditioning
Huanhuan Liu et al.
Biotechnology for biofuels, 12, 101-101 (2019-05-07)
Clostridium acetobutylicum is a model fermentative anaerobe for consolidated bioprocessing of lignocellulose hydrolysates into acetone-butanol-ethanol (ABE). However, the main inhibitors (acids, furans and phenols) ubiquitous in lignocellulose hydrolysates strictly limit the conversion efficiency. Thus, it is essential to understand the
Federica Murgia et al.
Reproductive sciences (Thousand Oaks, Calif.), 28(3), 728-735 (2020-11-12)
Endometriosis is a common disease affecting women in reproductive age. There are several hypotheses on the pathogenesis of this disease. Often, its lesions and symptoms overlap with those of many other medical and surgical conditions, causing a delay in diagnosis.
Tanya S McDonald et al.
Epilepsia, 58(7), 1172-1180 (2017-06-21)
To determine changes in glucose metabolism and the enzymes involved in the hippocampus ictally and postictally in the acute mouse flurothyl seizure model. [U- During seizures, total lactate levels increased 1.7-fold; however, [M + 3] enrichment of both lactate and
Judith Kouassi Nzoughet et al.
Metabolites, 10(2) (2020-02-06)
Glaucoma is an age related disease characterized by the progressive loss of retinal ganglion cells, which are the neurons that transduce the visual information from the retina to the brain. It is the leading cause of irreversible blindness worldwide. To

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service