Skip to Content
Merck
All Photos(2)

Key Documents

69455

Supelco

Chebulagic acid

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H30O27
CAS Number:
Molecular Weight:
954.66
UNSPSC Code:
85151701
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥85.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

SMILES string

O[C@H]1[C@H]2[C@H](CC(O)=O)C(=O)O[C@@H]3[C@H]4COC(=O)c5cc(O)c(O)c(O)c5-c6c(O)c(O)c(O)cc6C(=O)O[C@@H]3[C@@H](OC(=O)c7cc(O)c(O)c(OC1=O)c27)[C@@H](O4)OC(=O)c8cc(O)c(O)c(O)c8

InChI

1S/C41H30O27/c42-13-1-8(2-14(43)24(13)49)35(56)68-41-34-33-31(64-39(60)12(6-19(47)48)22-23-11(38(59)67-34)5-17(46)27(52)32(23)65-40(61)30(22)55)18(63-41)7-62-36(57)9-3-15(44)25(50)28(53)20(9)21-10(37(58)66-33)4-16(45)26(51)29(21)54/h1-5,12,18,22,30-31,33-34,41-46,49-55H,6-7H2,(H,47,48)/t12-,18+,22-,30-,31+,33-,34+,41-/m0/s1

InChI key

HGJXAVROWQLCTP-YABCKIEDSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mi-Jeong Ahn et al.
Planta medica, 68(5), 457-459 (2002-06-12)
The bioassay-directed isolation of Terminalia chebula fruits afforded four human immunodeficiency virus type 1 (HIV-1) integrase inhibitors, gallic acid ( 1) and three galloyl glucoses ( 2 - 4). In addition, four flavonol glycoside gallates ( 5 - 8) from
S M Hecht et al.
Journal of natural products, 55(4), 401-413 (1992-04-01)
While the design of molecules that inhibit or antagonize the functions of specific macromolecules is now well precedented, in many cases the structural information requisite to the design process is lacking. The tools of molecular biology can now furnish the
Sivasankar Shanmuganathan et al.
Vascular pharmacology, 108, 23-35 (2018-04-22)
Tumor necrosis factor-α (TNFα) a pleiotropic cytokine induces pro-inflammatory and pro-angiogenic changes in conditions such as diabetic retinopathy (DR) and neovascular age related macular degeneration (NV-AMD). Hence, inhibition of TNFα mediated changes can benefit the management of DR and NV-AMD.
Liang-Tzung Lin et al.
BMC microbiology, 13, 187-187 (2013-08-09)
We previously identified two hydrolyzable tannins, chebulagic acid (CHLA) and punicalagin (PUG) that blocked herpes simplex virus type 1 (HSV-1) entry and spread. These compounds inhibited viral glycoprotein interactions with cell surface glycosaminoglycans (GAGs). Based on this property, we evaluated
Ajay Kesharwani et al.
BMC complementary and alternative medicine, 17(1), 110-110 (2017-02-16)
Development of new and effective therapeutics for sexually transmitted herpes simplex virus-2 (HSV-2) infection is important from public health perspective. With an aim to identify natural products from medicinal plants, in the present study, the potential of Terminalia chebula Retz

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service