Skip to Content
Merck
All Photos(2)

Key Documents

61386

Supelco

Diosmin

analytical standard

Synonym(s):

3′,5,7-Trihydroxy-4′-methoxyflavone 7-rutinoside, Barosmin, Buchu resin, Diosmetin 7-O-rhamnosylglucoside, Diosmetin 7-rutinoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H32O15
CAS Number:
Molecular Weight:
608.54
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥90% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤10% water

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

COc1ccc(cc1O)C2=CC(=O)c3c(O)cc(O[C@@H]4O[C@H](CO[C@@H]5O[C@@H](C)[C@H](O)[C@@H](O)[C@H]5O)[C@@H](O)[C@H](O)[C@H]4O)cc3O2

InChI

1S/C28H32O15/c1-10-21(32)23(34)25(36)27(40-10)39-9-19-22(33)24(35)26(37)28(43-19)41-12-6-14(30)20-15(31)8-17(42-18(20)7-12)11-3-4-16(38-2)13(29)5-11/h3-8,10,19,21-30,32-37H,9H2,1-2H3/t10-,19+,21-,22+,23+,24-,25+,26+,27+,28+/m0/s1

InChI key

GZSOSUNBTXMUFQ-YFAPSIMESA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diosmin is a flavonoid glycoside that occurs naturally as an active constituent of citrus fruits.(1) It possess a wide range of pharmacological properties like antioxidant, anticarcinogenic, blood-lipid lowering and also anti-inflammatory activities. Diosmin can also find applications in the healing of venous ulcers, enhancement of microcirculation and improvement of venous tone.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Citrus flavonoid. Modification of hesperidin. Studied for its chemopreventive properties, including anti-proliferative and anti-inflammatory.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

This compound is commonly found in plants of the genus: mentha

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Jacqueline M Cohen et al.
Chest, 141(2), 308-320 (2012-02-09)
Postthrombotic syndrome (PTS) is a frequent, chronic complication of DVT. The effectiveness and safety of available treatments are unknown. The objective of this study was to systematically review the literature to assess whether pharmacologic and compression therapies are effective and
Simultaneous reversed-phase high-performance liquid chromatographic method for the determination of diosmin, hesperidin and naringin in different citrus fruit juices and pharmaceutical formulations
Kanaze IF, et al.
Journal of Pharmaceutical and Biomedical Analysis, 33, 243-249 (2003)
Mir Tahir et al.
Alcohol (Fayetteville, N.Y.), 47(2), 131-139 (2013-02-20)
The present investigation was designed to evaluate the efficacy of diosmin against ethanol-induced hepatotoxicity in rats by modulating various mechanisms including ethanol metabolizing enzymes, generation of free radicals, imbalance in oxidant-antioxidant status, oxidative damage to membrane lipids, activation of transcription
Tran Duc Dung et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 50(5), 1802-1810 (2012-02-01)
Hepatocellular carcinoma is a common type of cancer with poor prognosis. This study examines the in vitro and in vivo mechanisms of diosmin on human hepato-cellular carcinoma HA22T cell proliferation inhibition. HA22T cells were treated with different diosmin concentrations and
M Alcaraz et al.
Dento maxillo facial radiology, 40(5), 310-314 (2011-06-24)
The aim of this study was to evaluate the antioxidant substances present in the human diet with an antimutagenic protective capacity against genotoxic damage induced by exposure to X-rays in an attempt to reduce biological damage to as low a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service