Skip to Content
Merck
All Photos(1)

Key Documents

39441

Millipore

Nitrate Reagent B

suitable for microbiology

Synonym(s):

Sulfanilic acid solution, 4-Aminobenzenesulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41171621
PubChem Substance ID:
NACRES:
NA.85
Pricing and availability is not currently available.

shelf life

limited shelf life, expiry date on the label

Quality Level

composition

acetic acid (30%), 1000 mL
sulfanilic acid, 8.0 g

technique(s)

microbe id | metabolite detection: suitable

application(s)

agriculture
clinical testing
environmental
food and beverages
pharmaceutical

microbiology

suitability

Enterobacter spp.
Escherichia coli
Neisseria spp.
anaerobic bacteria
coliforms

SMILES string

Nc1ccc(cc1)S(O)(=O)=O

InChI

1S/C6H7NO3S/c7-5-1-3-6(4-2-5)11(8,9)10/h1-4H,7H2,(H,8,9,10)

InChI key

HVBSAKJJOYLTQU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

α-Naphthylamine and sulfanilic acid are used for detection of nitrate reduction by bacteria. Organisms containing nitrate reductase reduce nitrate to nitrite, which forms a diazonium salt with sulfanilic acid and reacts with α-naphthylamine to form a red azo dye.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Han Ming Gan et al.
Chemosphere, 82(4), 507-513 (2010-11-26)
A co-culture consisting of Hydrogenophaga sp. PBC and Ralstonia sp. PBA, isolated from textile wastewater treatment plant could tolerate up to 100 mM 4-aminobenzenesulfonate (4-ABS) and utilize it as sole carbon, nitrogen and sulfur source under aerobic condition. The biodegradation
Han Ming Gan et al.
Microbiology (Reading, England), 158(Pt 8), 1933-1941 (2012-05-23)
The gene coding for the oxygenase component, sadA, of 4-aminobenzenesulfonate (4-ABS) 3,4-dioxygenase in Hydrogenophaga sp. PBC was previously identified via transposon mutagenesis. Expression of wild-type sadA in trans restored the ability of the sadA mutant to grow on 4-ABS. The
Kwan Kim et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 100, 10-14 (2012-02-14)
We estimated the apparent size of the 'hot site' for surface-enhanced Raman scattering (SERS) located within the gaps between Pt nanoparticles and a flat Ag substrate. Initially, no Raman peaks were detected for 4-aminobenzenethiol (4-ABT) on a flat Ag substrate.
Han Ming Gan et al.
FEMS microbiology letters, 318(2), 108-114 (2011-02-18)
Genes involved in the 4-aminobenzenesulfonate (4-ABS) degradation pathway of Hydrogenophaga sp. PBC were identified using transposon mutagenesis. The screening of 10,000 mutants for incomplete 4-ABS biotransformation identified four mutants with single transposon insertion. Genes with insertions that impaired the ability
Ke-Jing Huang et al.
Colloids and surfaces. B, Biointerfaces, 61(2), 176-181 (2007-09-18)
A chemically modified electrode is constructed based on the multi-walled carbon nanotubes (MWNTs)/4-aminobenzeresulfonic acid (4-ABSA) film-coated glassy carbon electrode. The electrocatalytic oxidation of tyrosine (Tyr) is investigated on the surface of the MWNTs/4-ABSA-modified electrode using cyclic voltammetry (CV) and differential

Articles

Vibrios are motile, curved or comma-shaped bacilli and have a single polar flagella with sheet proteins. They are often found in open water, freshwater and saltwater.

There are many other methods of detection to indicate the presence of E. coli. Review common tests and biochemical reactions for this contaminant.

There are many other methods of detection to indicate the presence of E. coli. Review common tests and biochemical reactions for this contaminant.

There are many other methods of detection to indicate the presence of E. coli. Review common tests and biochemical reactions for this contaminant.

See All

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service