Skip to Content
Merck
All Photos(1)

Key Documents

86090

Sigma-Aldrich

Sulfanilic acid

puriss. p.a., ≥99.0% (T)

Synonym(s):

4-Aminobenzenesulfonic acid, Aniline-4-sulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
4-(H2N)C6H4SO3H
CAS Number:
Molecular Weight:
173.19
Beilstein:
908765
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

grade

puriss. p.a.

Quality Level

Assay

≥99.0% (T)

form

powder or crystals

impurities

≤1% ortho/meta-aminobenzene sulfonic acid (HPLC)

ign. residue

≤0.01% (as SO4)

mp

>300 °C (lit.)

anion traces

chloride (Cl-): ≤20 mg/kg
nitrite (NO2-): ≤0.5 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

SMILES string

Nc1ccc(cc1)S(O)(=O)=O

InChI

1S/C6H7NO3S/c7-5-1-3-6(4-2-5)11(8,9)10/h1-4H,7H2,(H,8,9,10)

InChI key

HVBSAKJJOYLTQU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Sulfanilic acid may be used for the functionalization of single-walled carbon nanotubes (SWNTs) with aryl sulfonate groups.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adsorption of essential micronutrients by carbon nanotubes and the implications for nanotoxicity testing.
Lin Guo et al.
Small (Weinheim an der Bergstrasse, Germany), 4(6), 721-727 (2008-05-28)
Kinetic, mechanistic and spectral studies for the oxidation of sulfanilic acid by alkaline hexacyanoferrate (III).
Farokhi SA and Nandibewoor ST.
Tetrahedron, 59(38), 7595-7602 (2003)
Starlee Lively et al.
Frontiers in cellular neuroscience, 12, 433-433 (2018-12-14)
Addressing potential sex differences in pre-clinical studies is crucial for developing therapeutic interventions. Although sex differences have been reported in epidemiological studies and from clinical experience, most pre-clinical studies of neuroinflammation use male rodents; however, sexual dimorphisms in microglia might
Nam Hoon Kim et al.
Nano letters, 10(10), 4181-4185 (2010-09-25)
A bifunctional adenosine-sensitive double-stranded DNA aptamer was used to create and control a surface-enhanced Raman spectroscopy (SERS) hot spot between a bulk Au surface and a gold nanoparticle (Au NP) attached to the aptamer via a biotin-avidin linkage. The Au
Prashant Barsing et al.
Bioresource technology, 102(2), 765-771 (2010-09-25)
A novel bacterial consortium (TJ-2) for mineralization of aromatic amines resulting from decolorization of azo dyes was developed. Three bacterial strains were identified as Pseudomonas pseudoalcaligenes (TJ-21,EU072476), Pseudomonas citronellolis (TJ-22,EU072477) and Pseudomonas testosterone (TJ-23,EU072477) by 16S rRNA gene sequence analysis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service