Skip to Content
Merck
All Photos(1)

Key Documents

38290

Sigma-Aldrich

Diisopropylamine

Diisopropylamine

puriss. p.a., ≥99.0% (GC)

Synonym(s):

DIPA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHNHCH(CH3)2
CAS Number:
Molecular Weight:
101.19
Beilstein:
605284
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥99.0% (GC)
bp:
84 °C (lit.)
vapor pressure:
50 mmHg ( 20 °C)
Pricing and availability is not currently available.

vapor density

3.5 (vs air)

Quality Level

vapor pressure

50 mmHg ( 20 °C)

grade

puriss. p.a.

Assay

≥99.0% (GC)

form

liquid

autoignition temp.

599 °F

expl. lim.

8.5 %

impurities

≤0.5% water

evapn. residue

≤0.001%

refractive index

n20/D 1.392 (lit.)
n20/D 1.393

bp

84 °C (lit.)

mp

−61 °C (lit.)

solubility

H2O: soluble 110 g/L at 25 °C (completely)

density

0.722 g/mL at 25 °C (lit.)

cation traces

Al: ≤0.5 mg/kg
Ba: ≤0.1 mg/kg
Bi: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.02 mg/kg
Fe: ≤0.5 mg/kg
K: ≤0.5 mg/kg
Li: ≤0.1 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.02 mg/kg
Mo: ≤0.1 mg/kg
Na: ≤0.5 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.1 mg/kg
Sr: ≤0.1 mg/kg
Zn: ≤0.1 mg/kg

SMILES string

CC(C)NC(C)C

InChI

1S/C6H15N/c1-5(2)7-6(3)4/h5-7H,1-4H3

InChI key

UAOMVDZJSHZZME-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Solvent extraction in environmental technology: Diisopropylamine was utilized in temperature swing solvent extraction for the zero liquid discharge of ultrahigh-salinity brines, demonstrating its utility in environmental cleanup and water treatment processes (Boo et al., 2020).

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

7.8 °F - closed cup

Flash Point(C)

-13.45 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Selective permeation of carbon dioxide through plasma polymerized membrane from diisopropylamine.
Matsuyama H, et al.
Journal of Membrane Science, 92(3), 257-265 (1994)
A L Kayushin et al.
Nucleic acids research, 24(19), 3748-3755 (1996-10-01)
Trinucleotide phosphoramidites that correspond to the codons of all 20 amino acids were synthesized in high yield in 5g scale. Precursors of those amidites--trinucleotide phosphotriesters--have been prepared using the phosphotriester approach without protection of the 3'-hydroxyl function. The structures of
Reaction of amines and acid catalysts. IV. Kinetics and mechanism of olefin formation from triethylamine and diisopropylamine on ?-alumina.
Hogan P and Pasek J.
Collection of Czechoslovak Chemical Communications, 38(5), 1513-1521 (1973)
Synthesis of high carbon materials from acetylenic precursors. Preparation of aromatic monomers bearing multiple ethynyl groups.
Neenan TX and Whitesides GM.
The Journal of Organic Chemistry, 53(11), 2489-2496 (1988)
Songping Liao et al.
Journal of the American Chemical Society, 125(49), 15114-15127 (2003-12-05)
Lithium diisopropylamide-mediated lithiations of N-alkyl ketimines derived from cyclohexanones reveal that simple substitutions on the N-alkyl side chain and the 2-position of the cyclohexyl moiety afford a 60,000-fold range of rates. Detailed rate studies implicate monosolvated monomers at the rate-limiting

Questions

Reviews

Active Filters

  1. 1 Ratings-only review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service