Merck
  • Lithium diisopropylamide-mediated lithiations of imines: insights into highly structure-dependent rates and selectivities.
Lithium diisopropylamide-mediated lithiations of imines: insights into highly structure-dependent rates and selectivities.
Journal of the American Chemical Society (2003-12-05)
Songping Liao, David B Collum
ABSTRACT
Lithium diisopropylamide-mediated lithiations of N-alkyl ketimines derived from cyclohexanones reveal that simple substitutions on the N-alkyl side chain and the 2-position of the cyclohexyl moiety afford a 60,000-fold range of rates. Detailed rate studies implicate monosolvated monomers at the rate-limiting transition structures in all instances. Comparisons of experimentally derived regioselectivities and rates, taken in conjunction with density functional theory computational studies, reveal a number of factors that influence reactivities including: (a) axial versus equatorial disposition of the proton on the cyclohexane ring, (b) syn versus anti orientation of the lithiation relative to the N-alkyl group, (c) the presence or absence of a potentially chelating methoxy moiety on the N-alkyl group, (d) the presence of a 2-methyl substituent at the geminal or distal alpha-carbon, and (e) branching in the N-alkyl group. The isolated contributions are not large, yet they display a strong and predictable additivity leading to a kinetic resolution of imines derived from racemic 2-methylcyclohexanone.
Sigma-Aldrich
Diisopropylamine, SAJ first grade, ≥98.0%
Sigma-Aldrich
Diisopropylamine, ≥99.5%
Sigma-Aldrich
Lithium diisopropylamide, 97%
Supelco
Diisopropylamine, analytical standard
Sigma-Aldrich
Diisopropylamine, purified by redistillation, 99.95%
Sigma-Aldrich
Lithium diisopropylamide solution, 2.0 M in THF/heptane/ethylbenzene
Sigma-Aldrich
Diisopropylamine, puriss. p.a., ≥99.0% (GC)