Skip to Content
Merck
All Photos(2)

Documents

45808

Supelco

Tricyclazole

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H7N3S
CAS Number:
Molecular Weight:
189.24
Beilstein:
980261
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

Cc1cccc2sc3nncn3c12

InChI

1S/C9H7N3S/c1-6-3-2-4-7-8(6)12-5-10-11-9(12)13-7/h2-5H,1H3

InChI key

DQJCHOQLCLEDLL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tricyclazole is a protectant, systemic fungicide, extensively used for the control of rice blast.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Tricyclazole may be used as an analytical reference standard for the quantification of the analyte in environmental samples using solid phase extrcation followed by liquid chromatography.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 3

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrea Kunova et al.
Pest management science, 69(2), 278-284 (2012-08-31)
Rice blast, caused by Magnaporthe oryzae B. Couch sp. nov., is one of the most destructive rice diseases worldwide, causing substantial yield losses every year. In Italy, its management is based mainly on the use of two fungicides, azoxystrobin and
Seul Ah Jeong et al.
Bulletin of environmental contamination and toxicology, 89(6), 1181-1185 (2012-09-28)
Soil is the major sink for majority of pesticides applied on agricultural crops and its fate depends on variety of factors. There is little research on fate of pesticide in field soil under different climatic conditions and there is a
D Mares et al.
Mycological research, 110(Pt 6), 686-696 (2006-06-14)
Derivatives of pyrazolo[1,5-a][1, 3, 5]triazine-2,4-dione,pyrazolo[1,5-c][1, 3, 5]thiadiazine-2-one, pyrazolo[3,4-d][1, 3]thiazine-4-one, and pyrazolo[3,4-d][1, 3]thiazine-4-thione were screened for antifungal activity against the causal agent of rice blast disease, Magnaporthe grisea. The compounds were tested at doses ranging from 10 to 200mugml(-1), using the
Marcel M L Cunha et al.
BMC microbiology, 10, 80-80 (2010-03-18)
The pathogenic fungus Fonsecaea pedrosoi constitutively produces the pigment melanin, an important virulence factor in fungi. Melanin is incorporated in the cell wall structure and provides chemical and physical protection for the fungus.We evaluated the production of nitric oxide (NO)
Rodrigo Almeida-Paes et al.
Microbes and infection, 11(5), 554-562 (2009-03-31)
Sporothrix schenckii is known to produce DHN melanin on both conidial and yeast cells, however little information is available regarding the factors inducing fungal melanization. We evaluated whether culture conditions influenced melanization of 25 Brazilian S. schenckii strains and one

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service