Skip to Content
Merck
All Photos(1)

Key Documents

36967

Supelco

1,1-Dichloroethane

analytical standard, contains 100 ppm hydroquinone as stabilizer

Synonym(s):

Ethylidene chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CHCl2
CAS Number:
Molecular Weight:
98.96
Beilstein:
1696901
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

contains

100 ppm hydroquinone as stabilizer

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

neat

SMILES string

CC(Cl)Cl

InChI

1S/C2H4Cl2/c1-2(3)4/h2H,1H3

InChI key

SCYULBFZEHDVBN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1,1-Dichloroethane is typically utilized as an intermediate for the synthesis of vinyl chloride.

Application

1,1-Dichloroethane may be used as an analytical reference standard for the determination of the analyte in water and ambient air samples by gas chromatography (GC) based techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

14.0 °F - closed cup

Flash Point(C)

-10.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The determination of volatile organic compounds from EPA Method 524.2 using purge-and-trap capillary gas chromatography, ECD, and FID
Kessels H, et al.
Journal of Chromatographic Science, 30(7), 247-255 (1992)
Multicomponent analysis of volatile organic compounds in water by automated purge and trap coupled to gas chromatography-mass spectrometry
Martinez E, et al.
Journal of Chromatography A, 959(1-2), 181-190 (2002)
Multi-adsorbent preconcentration and gas chromatographic analysis of air toxics with an automated collection/analytical system
Pollack AJ, et al.
Journal of the Air & Waste Management Association (1995), 41(9), 1213-1217 (1991)
Halogenated Hydrocarbons
Comprehensive Toxicology, 7(9), 459-474 (2010)
Lewis Semprini et al.
Journal of contaminant hydrology, 103(3-4), 157-167 (2008-11-22)
A field study was performed to evaluate the potential for in-situ aerobic cometabolism of 1,1,1-trichloroethane (1,1,1-TCA) through bioaugmentation with a butane enrichment culture containing predominantly two Rhodococcus sp. strains named 179BP and 183BP that could cometabolize 1,1,1-TCA and 1,1-dicholoroethene (1,1-DCE).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service