Skip to Content
Merck
All Photos(2)

Key Documents

36666

Supelco

Aldrin

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H8Cl6
CAS Number:
Molecular Weight:
364.91
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

ClC1=C(Cl)C2(Cl)C3C4CC(C=C4)C3C1(Cl)C2(Cl)Cl

InChI

1S/C12H8Cl6/c13-8-9(14)11(16)7-5-2-1-4(3-5)6(7)10(8,15)12(11,17)18/h1-2,4-7H,3H2/t4-,5+,6+,7-,10+,11-

InChI key

QBYJBZPUGVGKQQ-SJJAEHHWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Aldrin is a chlorinated cyclodiene[1][2], which is a synthetically produced pyrethroid insecticide.[3]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - STOT RE 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Organochlorine insecticide residues in drinking and ground water in and around Delhi
Mukherjee I and Gopal M
Environmental Monitoring and Assessment, 76, 185-193 (2002)
paper-based thioglycolic acid (TGA)-capped CdTe QD device for rapid screening of organophosphorus and carbamate insecticides
Apilux.A, et al.
Analytical Methods : Advancing Methods and Applications, 9, 519-527 (2017)
Long-term health effects of aldrin and dieldrin: a study of exposure, health effects and mortality of workers engaged in the manufacture and formulation of the insecticides aldrin and dieldrin
Jong DC
Toxicology Letters, 1-206 (1991)
Hans Ragnar Norli et al.
Journal of chromatography. A, 1218(41), 7234-7241 (2011-09-09)
The QuEChERS method developed for 22 organochlorine pesticides (OCPs) and 7-polychlorinated biphenyls (PCBs) in fish tissue involves a simple and efficient freezing technique for removal of lipids. The equipment developed consists of disposable syringes and a freezing block constructed from
A K Chopra et al.
Environmental monitoring and assessment, 173(1-4), 905-916 (2010-03-23)
In recent years, various environmental issues have aroused a concern on the pollution of pesticides in rivers and in their various intercompartments. Multiple residues of pesticides discharged from industries or as a result of extensive use of agrochemicals in agriculture

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service