Skip to Content
Merck
All Photos(1)

Key Documents

35487

Supelco

4,4′-DDE

PESTANAL®, analytical standard

Synonym(s):

1,1-Dichloro-2,2-bis(4-chlorophenyl)ethene, 4,4′-DDE

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(ClC6H4)2C=CCl2
CAS Number:
Molecular Weight:
318.03
Beilstein:
1913355
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

88-90 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

Clc1ccc(cc1)\C(=C(/Cl)Cl)c2ccc(Cl)cc2

InChI

1S/C14H8Cl4/c15-11-5-1-9(2-6-11)13(14(17)18)10-3-7-12(16)8-4-10/h1-8H

InChI key

UCNVFOCBFJOQAL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4,4′-DDE can be found to be a breakdown product of trichloro-2,2-bis(4-chlorophenyl)ethane, in an aerobic environment. It can behave like a typical potent androgen receptor antagonist.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - STOT RE 1 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cometabolism of 1, 1-dichloro-2, 2-bis (4-chlorophenyl) ethylene by Pseudomonas acidovorans M3GY grown on biphenyl
Hay.GA and Focht.DD
Applied and Environmental Microbiology, 64(6), 2141- 2146 (1998)
Metabolism of 1, 1-dichloro-2, 2-bis (4-chlorophenyl) ethylene by Alcaligenes denitrificans
Ahuja R, et al.
Biotechnology Letters, 23(6), 423- 426 (2001)
Anita H Poulsen et al.
Environmental pollution (Barking, Essex : 1987), 175, 92-99 (2013-01-29)
This study evaluated the dietary uptake kinetics and sublethal toxicity of p,p'-dichlorodiphenyl dichloroethylene (p,p'-DDE) in Antarctic krill. The uptake rate constant (characterised by the seawater volume stripped of contaminant sorbed to algae) of 200 ± 0.32 mL g(-1) wet weight
Roberto De La Torre-Roche et al.
Environmental science & technology, 47(2), 718-725 (2012-12-21)
The effect of nanoparticle (NP), bulk, or ionic Ag exposure on dichlorodiphenyldichloroethylene (p,p'-DDE; DDT metabolite) accumulation by Glycine max L. (soybean) and Cucurbita pepo L. (zucchini) was investigated. The plants were grown in 125-mL jars of vermiculite amended with 500
Immle Delvaux et al.
Environmental research, 132, 24-32 (2014-04-20)
The study aim was to investigate the association between prenatal exposure to endocrine disrupting chemicals (EDCs) and the body composition of 7 to 9 year old Flemish children. The subjects were 114 Flemish children (50% boys) that took part in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service