Skip to Content
Merck
All Photos(1)

Key Documents

36129

Supelco

Hexazinone

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H20N4O2
CAS Number:
Molecular Weight:
252.31
Beilstein:
618801
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CN(C)C1=NC(=O)N(C2CCCCC2)C(=O)N1C

InChI

1S/C12H20N4O2/c1-14(2)10-13-11(17)16(12(18)15(10)3)9-7-5-4-6-8-9/h9H,4-8H2,1-3H3

InChI key

CAWXEEYDBZRFPE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hexazinone is a systemic triazine herbicide, which can find applications in weed control management, via inhibition of photosynthesis in the target plants.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Tatiane Severo Silva et al.
Chemosphere, 236, 124333-124333 (2019-07-19)
The use of herbicides in Brazil has been carried out based on the manufacturer's recommendation, often disregarding the high variability of soil attributes. The use of statistical methods to predict the herbicide retention processes in the soil can contribute to
S Stone et al.
Environmental toxicology and chemistry, 40(2), 473-486 (2020-11-18)
Agriculture within the Great Barrier Reef catchment area has contributed to pesticide contamination of adjacent freshwater ecosystems that flow into the Great Barrier Reef World Heritage Area. A novel multispecies toxicity test was used to assess the toxicity of diuron
Guangyang Liu et al.
Sensors (Basel, Switzerland), 18(5) (2018-05-19)
Various highly sensitive and selective analytical methods have been used to monitor metsulfuron-methyl residue in the environment. However, these methods involve costly instruments and complex, time-consuming operations performed in laboratories. Here, a rapid, convenient, and sensitive colorimetric sensor based on
Coastal Plain and Piedmont, Vegetation Management: Environmental Impact Statement, 2 (1989)
W L Strong et al.
Journal of environmental management, 77(3), 194-204 (2005-08-17)
A soil-active herbicide (hexazinone) was applied (0, 2, and 4 kg/ha of active ingredient) in a 3-year-old regenerating boreal Populus tremuloides Michx. (aspen) clearcut to determine its effect on the compositional and structural development of the vegetation. Woody stem densities

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service