Skip to Content
Merck
All Photos(1)

Documents

33996

Supelco

Ethopabate

VETRANAL®, analytical standard

Synonym(s):

Methyl 4-acetamido-2-ethoxybenzoate, Ethopabate, Ethyl pabate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H15NO4
CAS Number:
Molecular Weight:
237.25
Beilstein:
2808914
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

CCOc1cc(NC(C)=O)ccc1C(=O)OC

InChI

1S/C12H15NO4/c1-4-17-11-7-9(13-8(2)14)5-6-10(11)12(15)16-3/h5-7H,4H2,1-3H3,(H,13,14)

InChI key

GOVWOKSKFSBNGD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethopabate is an antibiotic used as a coccidiostat in poultry feeds.

Application

Ethopabate may be used as a reference standard in the determination of ethopabate in feed samples using solid phase extraction with high performance liquid chromatography (SPE-HPLC).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The treatment of coccidiosis: studies on the sensitivity of recent field isolates of Eimeria acervulina type to anticoccidial drugs given in the drinking water.
H D Chapman
Journal of comparative pathology, 92(2), 213-218 (1982-04-01)
Illegal use of Amprol-Plus for game.
O Swarbrick
The Veterinary record, 127(12), 315-315 (1990-09-22)
H D Chapman
Research in veterinary science, 46(3), 419-420 (1989-05-01)
Treatment with toltrazuril, sulphaquinoxaline/pyrimethamine and amprolium/ethopabate prevented mortality in chickens infected with field isolates of Eimeria tenella. Amprolium/ethopabate was the most effective drug in reducing lesions caused by the parasites. Few oocysts of E tenella were produced in birds medicated
A C Voeten et al.
Tijdschrift voor diergeneeskunde, 103(23), 1284-1289 (1978-12-01)
A floor pen trial was carried out on broilers in 1975 to compare the anticoccidial efficacy of monensin1), 3,5 dinitro-o-toluamide2), clopidol3), amprolium + ethopabacte4), amprolium + ethopabate + sulfaquinoxaline + pyrimethamine5) and robenidine6) in the presence of untreated controls. 4,200
C A Kan et al.
Tijdschrift voor diergeneeskunde, 114(2), 76-82 (1989-01-15)
Amprolium may be used as a coccidiostat in rearing hens and is a therapeutical agent used in laying hens. As a result of cross contamination, low amprolium levels may occur in feed. Feed containing a concentration of amprolium ranging from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service