Skip to Content
Merck
All Photos(1)

Key Documents

33931

Supelco

Clioquinol

VETRANAL®, analytical standard

Synonym(s):

5-Chloro-7-iodo-8-quinolinol, 5-Chloro-8-hydroxy-7-iodoquinoline, Clioquinol, Iodochlorhydroxyquin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H5ClINO
CAS Number:
Molecular Weight:
305.50
Beilstein:
153637
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Oc1c(I)cc(Cl)c2cccnc12

InChI

1S/C9H5ClINO/c10-6-4-7(11)9(13)8-5(6)2-1-3-12-8/h1-4,13H

InChI key

QCDFBFJGMNKBDO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Clioquinol is a 8-hydroxyquinoline derivative, which comes under the category of allergens and finds potential applications as an anti-infective agent for diarrhoea. It can be used like a chelator in the treatment of alzheimer′s disease, since it has good affinity towards zinc and copper ions and can easily cross the blood-brain barrier.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Clioquinol, a drug for Alzheimer's disease specifically interfering with brain metal metabolism: structural characterization of its zinc (II) and copper (II) complexes
Vaira DM, et al
Inorganic Chemistry, 43, 3795-3379 (2004)
Treatment of Alzheimer?s disease with clioquinol
Regland B, et al.
Dementia and Geriatric Cognitive Disorders, 12, 408-414 (2001)
Simon Melov
Trends in neurosciences, 25(3), 121-123 (2002-02-20)
Alzheimer's disease (AD) is a devastating age-related neurodegenerative disorder that has been intensively studied over the last several years. In vitro and in vivo studies have led to an understanding of some of the physico-chemical properties of amyloid, a well-characterized
Lin W Hung et al.
Future medicinal chemistry, 4(8), 955-969 (2012-06-02)
In 1906, Alois Alzheimer first characterized the disease that bears his name. Despite intensive research, which has led to a better understanding of the pathology, there is no effective treatment for this disease. Of the drugs approved by the US
Peter J Crouch et al.
Journal of neurochemistry, 119(1), 220-230 (2011-07-30)
Impaired metal ion homeostasis causes synaptic dysfunction and treatments for Alzheimer's disease (AD) that target metal ions have therefore been developed. The leading compound in this class of therapeutic, PBT2, improved cognition in a clinical trial with AD patients. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service