Skip to Content
Merck
All Photos(2)

Key Documents

D123609

Sigma-Aldrich

5,7-Diiodo-8-hydroxyquinoline

95%

Synonym(s):

5,7-Diiodo-8-quinolinol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H5I2NO
CAS Number:
Molecular Weight:
396.95
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

crystals

mp

>200 °C (dec.) (lit.)

SMILES string

Oc1c(I)cc(I)c2cccnc12

InChI

1S/C9H5I2NO/c10-6-4-7(11)9(13)8-5(6)2-1-3-12-8/h1-4,13H

InChI key

UXZFQZANDVDGMM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

American Academy of Pediatrics Committee on Drugs: Clioquinol (iodochlorhydroxyquin, vioform) and iodoquinol (diiodohydroxyquin): blindness and neuropathy.
Pediatrics, 86(5), 797-798 (1990-11-01)
M T González-Garza et al.
Archivos de investigacion medica, 20(1), 29-32 (1989-01-01)
Monophasic liquid media for axenical cultivation of Entamoeba histolytica, have permitted to define diverse aspects of this parasite's biology, without associated organisms interference. In addition, these media have allowed the in vitro evaluation of new drugs with potential antiamebic properties.
Spectrophotometric determination of clioquinol and diodoquin in pharmaceutical preparations using uranyl acetate as a chromogenic agent.
U Saha et al.
The Analyst, 113(11), 1653-1655 (1988-11-01)
Shih-Bin Su et al.
Infection control and hospital epidemiology, 28(6), 679-683 (2007-05-24)
Investigation of a program to eradicate amebiasis using consecutive intensive mass screenings followed by medication in a large institute for adults with mental retardation in Taiwan. Prospective cohort study, with 3 years of follow-up. A large, 450-bed institution for adults
R J Citronberg et al.
Clinical infectious diseases : an official publication of the Infectious Diseases Society of America, 20(3), 700-702 (1995-03-01)
Genital amebiasis is a rare complication of infection with Entamoeba histolytica, even in areas where the pathogen is endemic. We describe a patient who apparently contracted intestinal amebiasis on a trip to Mexico and who presented with ulcerative vulvovaginitis 2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service