Skip to Content
Merck
All Photos(1)

Documents

32419

Supelco

Novaluron

PESTANAL®, analytical standard

Synonym(s):

1-[3-Chloro-4-(1,1,2-trifluoro-2-trifluoromethoxyethoxy)phenyl]-3-(2,6-difluorobenzoyl)urea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H9ClF8N2O4
CAS Number:
Molecular Weight:
492.70
Beilstein:
5465888
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

FC(OC(F)(F)F)C(F)(F)Oc1ccc(NC(=O)NC(=O)c2c(F)cccc2F)cc1Cl

InChI

1S/C17H9ClF8N2O4/c18-8-6-7(4-5-11(8)31-16(22,23)14(21)32-17(24,25)26)27-15(30)28-13(29)12-9(19)2-1-3-10(12)20/h1-6,14H,(H2,27,28,29,30)

InChI key

NJPPVKZQTLUDBO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Novaluron (Rimon) is a type of benzoylphenyl urea,3 that acts as an insect growth regulator by inhibiting chitin synthesis. It acts as a powerful toxicant for controlling lepidopteran larvae. It is suitable in the rainy and tropics seasons, as it is a rainfast compound.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novaluron (Rimon), a novel IGR: Potency and cross-resistance.
Ishaaya, Isaac, Svetlana Kontsedalov, and A. Rami Horowitz.
Archives of Insect Biochemistry and Physiology, 54.4, 157-164 (2003)
Laboratory and field evaluation of novaluron, a new acylurea insect growth regulator, against Aedes aegypti (Diptera: Culicidae).
Mulla, Mir S., et al.
Journal of Vector Ecology : Journal of the Society for Vector Ecology, 28, 241-254 (2003)
Gayan P Withanage et al.
BioMed research international, 2020, 9567019-9567019 (2020-03-20)
Dengue is the most important mosquito-borne viral infection in Sri Lanka causing an enormous social and economic burden in the country. In the absence of therapeutic drugs and the developed vaccines are under investigation, vector control is the best strategy
Novaluron (Rimon), a novel IGR--mechanism, selectivity and importance in IPM programs.
Ishaaya, I., et al.
Mededelingen (Rijksuniversiteit te Gent. Fakulteit Van de Landbouwkundige en Toegepaste Biologische Wetenschappen), 67.3, 617-626 (2001)
Abid Hussain et al.
Molecules (Basel, Switzerland), 24(23) (2019-11-30)
Disruption in chitin regulation by using chitin synthesis inhibitor (novaluron) was investigated to gain insights into the biological activity of chitinase in red palm weevils, an invasive pest of date palms in the Middle East. Impact of novaluron against ninth

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service