Skip to Content
Merck
All Photos(1)

Documents

31730

Sigma-Aldrich

Diethylamine

puriss. p.a., ≥99.5% (GC)

Synonym(s):

N,N-Diethylamine, N-ethyl-ethanamine, DEA, Diethamine, Diethyl amine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)2NH
CAS Number:
Molecular Weight:
73.14
Beilstein:
605268
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

vapor density

2.5 (vs air)

Quality Level

vapor pressure

14.14 psi ( 55 °C)
180 mmHg ( 20 °C)
3.51 psi ( 20 °C)

grade

puriss. p.a.

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

594 °F

expl. lim.

10.1 %

impurities

≤0.1% water

evapn. residue

≤0.001%

refractive index

n20/D 1.385 (lit.)
n20/D 1.386

bp

55 °C (lit.)

mp

−50 °C (lit.)

density

0.707 g/mL at 25 °C (lit.)

cation traces

Al: ≤0.5 mg/kg
Ba: ≤0.1 mg/kg
Bi: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.05 mg/kg
Co: ≤0.02 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.02 mg/kg
Fe: ≤0.1 mg/kg
K: ≤0.5 mg/kg
Li: ≤0.1 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.02 mg/kg
Mo: ≤0.1 mg/kg
Na: ≤0.5 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.1 mg/kg
Sr: ≤0.1 mg/kg
Zn: ≤0.1 mg/kg

SMILES string

CCNCC

InChI

1S/C4H11N/c1-3-5-4-2/h5H,3-4H2,1-2H3

InChI key

HPNMFZURTQLUMO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Diethylamine is a secondary amine that is used in the production of pesticides, dyes, rubber, resins, and pharmaceuticals as a solvent, catalyst, and corrosion inhibitor.

Application

Diethylamine can be used as:
  • A catalyst in the tandem Knoevenagel-Michael reaction of dimedone with aromatic aldehydes to synthesize bis-dimedone derivatives.
  • A solvent to synthesize 4-quinolones via Sonogashira cross-coupling of iodoanilines and alkynes.
  • A base in the desymmetrizing cyclization of benzylic diallyl iodide to synthesize methylene indanes via intramolecular Heck reaction.
  • A template in the synthesis of silicoaluminophosphate (SAPO) molecular sieves.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-14.8 °F - closed cup

Flash Point(C)

-26 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Eagleson M.
Concise Encyclopedia Chemistry, 387-387 (1994)
Samira Mubareka et al.
Journal of clinical microbiology, 45(1), 231-233 (2006-11-10)
We compared the performances of five commercial rubella virus immunoglobulin G (IgG) avidity assays. The Adaltis (kappa = 0.28) and Diesse (kappa = 0.33) assays showed poor correlation, the Behring assay (kappa = 0.68) showed good correlation, and the Euroimmun
Lakshmi Thirumangalakudi et al.
Journal of neurochemistry, 106(1), 475-485 (2008-04-16)
Recent findings suggest that hypercholesterolemia may contribute to the onset of Alzheimer's disease-like dementia but the underlying mechanisms remain unknown. In this study, we evaluated the cognitive performance in rodent models of hypercholesterolemia in relation to neuroinflammatory changes and amyloid
Rakesh Naduvile Veedu et al.
The journal of physical chemistry. A, 112(40), 9742-9750 (2008-09-16)
Five aryliminopropadienones 4a- d have been synthesized by flash vacuum thermolysis (FVT) by using two different precursors in each case. These compounds were deposited at 50 K at a pressure of ca. 10(-6) mbar together with three different nucleophiles, namely
Jun Guo et al.
Journal of chromatography. A, 1307, 66-72 (2013-08-15)
The extraction efficiency of analytes in a cold fiber headspace solid-phase microextraction (CF-HS-SPME) system was investigated both theoretically and experimentally. The system was applied for quantitative extraction of polycyclic aromatic hydrocarbons (PAHs) from solid matrices. In order to achieve better

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service