Skip to Content
Merck
All Photos(1)

Key Documents

31659

Supelco

o-Tolidine

analytical standard

Synonym(s):

3,3′-Dimethylbenzidine, 4,4′-Bianisidine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-C6H3(CH3)-4-NH2]2
CAS Number:
Molecular Weight:
212.29
Beilstein:
2210640
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

128-131 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

Cc1cc(ccc1N)-c2ccc(N)c(C)c2

InChI

1S/C14H16N2/c1-9-7-11(3-5-13(9)15)12-4-6-14(16)10(2)8-12/h3-8H,15-16H2,1-2H3

InChI key

NUIURNJTPRWVAP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

o-Tolidine is widely used in the aniline dye industry for the synthesis of different azo dyes.

Application

o-Tolidine (3,3′-dimethylbenzidine) may be used as an analytical reference standard for the determination of the analyte formed from azo colorants in toy products by high-performance liquid chromatography with UV detection.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Sensitive colorimetric reagent for gold and for free chlorine in water.

Warning

May be carcinogenic.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Carcinogenic properties of orthotolidine (3, 3?-dimethylbenzidine).
Pliss GB and Zabezhinsky MA
Journal of the National Cancer Institute, 45(2), 283- 295 (1970)
Shinpei Kawarai et al.
The Journal of veterinary medical science, 72(2), 131-140 (2009-11-27)
It is essential to develop a technique to culture purified skin-derived mast cells (SMCs) to facilitate immunological research on allergic diseases in dogs. This study was performed to develop an efficient culture system for canine SMCs and to characterize the
Dyes metabolized to 3,3'-dimethylbenzidine (3,3'-dimethylbenzidine dye class.
Report on carcinogens : carcinogen profiles, 10, 105-106 (2004-08-25)
Zhijun Cao et al.
Biosensors & bioelectronics, 23(3), 348-354 (2007-06-06)
The electrooxidation of o-tolidine (oTD) was investigated via the electrochemical quartz crystal microbalance (EQCM) technique. The formation and breakage of the poorly soluble charge-transfer complex (CTC) occurred during the redox switching of oTD, and the CTC precipitation on and its
3,3'-Dimethylbenzidine and dyes metabolized to 3,3'-dimethylbenzidine: 3,3'-dimethylbenzidine.
Report on carcinogens : carcinogen profiles, 12, 168-170 (2011-08-20)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service