Skip to Content
Merck
All Photos(1)

Key Documents

48525

Supelco

3,3′-Dichlorobenzidine

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H10Cl2N2
CAS Number:
Molecular Weight:
253.13
EC Number:
UNSPSC Code:
12000000

grade

analytical standard

CofA

current certificate can be downloaded

packaging

ampule of 100 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

storage temp.

2-30°C

InChI

1S/C12H10Cl2N2/c13-9-5-7(1-3-11(9)15)8-2-4-12(16)10(14)6-8/h1-6H,15-16H2

InChI key

HUWXDEQWWKGHRV-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I Zwirner-Baier et al.
Archives of toxicology, 68(1), 8-14 (1994-01-01)
The release and availability of the carcinogenic component of the soluble azo dye Direct Red 46 and the insoluble azo pigment Pigment Yellow 17 were analyzed in Wistar rats using hemoglobin adducts as a dosimeter. The levels of hemoglobin adducts
Kristian F Knoell et al.
Journal of toxicology and environmental health. Part A, 75(8-10), 551-556 (2012-06-13)
In a chemical plant in Germany producing 3,3'-dichlorobenzidine dihydrochloride for the manufacture of colorants, blood and urine samples were taken for biological monitoring. 3,3'-Dichlorobenzidine (DBZ) was analyzed in urine by thin-layer chromatography and subsequently further combined with analysis of adducts
M C Nyman et al.
Journal of the American Society for Mass Spectrometry, 10(11), 1152-1156 (1999-10-28)
3,3'-Dichlorobenzidine (DCB) and its degradation products, 3-chlorobenzidine (MCB) and benzidine, are of environmental concern because of their carcinogenic nature. The suitability of a small Fourier transform ion cyclotron resonance (FT-ICR) mass spectrometer for the analysis of these environmental contaminants in
R Joppich-Kuhn et al.
International archives of occupational and environmental health, 69(4), 240-246 (1997-01-01)
A method based on gas chromatography/mass spectrometry-negative ion chemical ionization detection (GC/MS-NCI) was developed for the determination of 3,3'-dichlorobenzidine (DCB)-hemoglobin adducts. Adducts were released from hemoglobin by mild alkaline hydrolysis and determined by GC/MS-NCI after extraction and derivatization with heptafluorobutyric
Jin-Heon Lee et al.
Industrial health, 41(3), 242-248 (2003-08-15)
3,3'-dichlorobenzidine (DCB) is suspected to be arcinogenic in experimental animal and human. Several studies have investigated excretion of metabolites in urine, hemoglobin adduction and cancer incidence among workers exposed occupationally to DCB. In these researches, metabolites of DCB had a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service