Skip to Content
Merck
All Photos(2)

Key Documents

240834

Sigma-Aldrich

Diphenyl ether

ReagentPlus®, ≥99%

Synonym(s):

Diphenyl oxide, Phenyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2O
CAS Number:
Molecular Weight:
170.21
Beilstein:
1364620
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

>5.86 (25 °C, vs air)

vapor pressure

<1 mmHg ( 20 °C)

product line

ReagentPlus®

Assay

≥99%

form

crystalline

autoignition temp.

1144 °F

expl. lim.

1.5 %

refractive index

n20/D 1.579 (lit.)

bp

259 °C (lit.)

mp

25-27 °C (lit.)

solubility

alcohol: soluble(lit.)
benzene: soluble(lit.)
diethyl ether: soluble(lit.)
glacial acetic acid: soluble(lit.)
water: insoluble(lit.)

density

1.073 g/mL at 25 °C (lit.)

SMILES string

O(c1ccccc1)c2ccccc2

InChI

1S/C12H10O/c1-3-7-11(8-4-1)13-12-9-5-2-6-10-12/h1-10H

InChI key

USIUVYZYUHIAEV-UHFFFAOYSA-N

Gene Information

human ... TTR(7276)

Looking for similar products? Visit Product Comparison Guide

General description

The diphenyl derivatives, having potency for inhibiting InhA (an essential enoyl acyl carrier protein reductase involved in mycolic acid biosynthesis), were studied.

Application

Diphenyl ether can be used as:
  • An additive in the fabrication of thienothiophene-co-benzodithiophene polymer-based organic photovoltaics and isoindigo polymer-based organic solar cells.
  • A model compound to study the hydrodeoxygenation of ether linkages in the lignin fragments using various catalytic systems.
  • A solvent in the synthesis of FePd nanoparticles using modified polyol process.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

239.0 °F - closed cup

Flash Point(C)

115 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Kamsri et al.
SAR and QSAR in environmental research, 25(6), 473-488 (2014-05-03)
A series of diphenyl ether derivatives were developed and showed promising potency for inhibiting InhA, an essential enoyl acyl carrier protein reductase involved in mycolic acid biosynthesis, leading to the lysis of Mycobacterium tuberculosis. To understand the structural basis of
Bifunctional Ni catalysts for the one-pot conversion of Organosolv lignin into cycloalkanes
Wang X and Rinaldi R
Catalysis Today, 269, 48-55 (2016)
Transformation to L10 structure in FePd nanoparticles synthesized by modified polyol process
Watanabe K, et al.
Science and Technology of Advanced Materials, 7, 145-145 (2006)
Toward efficient thick active PTB7 photovoltaic layers using diphenyl ether as a solvent additive
Zheng Y, et al.
ACS Applied Materials & Interfaces, 8, 15724-15731 (2016)
Hydrodeoxygenation of lignin-derived phenolic compounds over bi-functional Ru/H-Beta under mild conditions
Yao G, et al.
Fuel: The Science and Technology of Fuel and Energy, 150, 175-183 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service