Skip to Content
Merck
All Photos(5)

Key Documents

493546

Sigma-Aldrich

Ethyl alcohol, Pure

greener alternative

≥99.50%, ACS reagent (Traceable to ACS product listing.), 200 proof, meets USP testing specifications

Synonym(s):

Ethanol, absolute alcohol, non-denatured ethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2OH
CAS Number:
Molecular Weight:
46.07
EC Number:
MDL number:
UNSPSC Code:
15101802
PubChem Substance ID:
NACRES:
NA.05
grade:
ACS reagent (Traceable to ACS product listing.)
Assay:
≥99.50%
bp:
78.3 °C
Pricing and availability is not currently available.

Product Name

Ethyl alcohol, Pure, 200 proof, meets USP testing specifications

grade

ACS reagent (Traceable to ACS product listing.)

Quality Level

Agency

meets USP testing specifications

vapor density

1.59 (vs air)

Assay

≥99.50%

form

liquid

expl. lim.

3.1-27.7 %

greener alternative product characteristics

Safer Solvents and Auxiliaries
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

200 proof

evapn. residue

≤0.0025%

pH

7.0 (20 °C, 10 g/L)

bp

78.3 °C

mp

-114 °C

solubility

water: soluble

λ

in H2O

UV absorption

λ: 240 nm Amax: 0.40
λ: 250 nm Amax: 0.30
λ: 260 nm Amax: 0.30
λ: 270 nm Amax: 0.10
λ: 340 nm Amax: 0.10

greener alternative category

SMILES string

OCC

InChI

1S/C2H6O/c1-2-3/h3H,2H2,1H3

InChI key

LFQSCWFLJHTTHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Prices include Federal Tax.
The product is a non-denatured 200proof ethanol. Ethanol, a primary alcohol, is an important raw material for the synthesis of various compounds. It can be synthesized either from molasses by fermentation or by hydration of ethylene. It acts as a hydrogen source and reaction medium during the transfer hydrogenation of carbonyl compounds in the presence of sodium hydroxide.[1] The impact of ethanol/water fumigation on the performance of diesel engines has been studied.[2]
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. Ethanol is environmentally preferable green solvent and thus it aligns with "Safer Solvents and Auxiliaries" and "Use of Renewable Feedstocks". 

Application

Pure ethyl alcohol has been used for the homogenization of tissue samples.[3]

Other Notes

Sales restrictions may apply

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benjamin M Pri-Tal et al.
General and comparative endocrinology, 174(2), 166-174 (2011-09-13)
The annual killifish, Austrofundulus limnaeus, typically enters embryonic diapause at two distinct points of development, termed diapause II and III. This study explores the role of maternal and embryonic steroid hormones, including 17-β-estradiol (E2), androstenedione (A4) and testosterone (T), in
Sodium hydroxide-catalyzed transfer hydrogenation of carbonyl compounds and nitroarenes using ethanol or isopropanol as both solvent and hydrogen donor.
Wang D, et al.
J. Mol. Catal. A: Chem., 400, 14-21 (2015)
Assessment of a direct injection diesel engine fumigated with ethanol/water mixtures.
Morsy MH.
Energy Conversion and Management , 94, 406-414 (2015)
Ross B Mounsey et al.
Experimental neurology, 273, 36-44 (2015-08-06)
Parkinson's disease (PD) is a common chronic neurodegenerative disorder, usually of idiopathic origin. Symptoms including tremor, bradykinesia, rigidity and postural instability are caused by the progressive loss of dopaminergic neurons in the nigrostriatal region of the brain. Symptomatic therapies are
S Enocksson et al.
The Journal of clinical investigation, 95(5), 2239-2245 (1995-05-01)
Although it is well established in several mammalian species that beta 3-adrenoceptors play a major role in regulating lipolysis and thermogenesis in adipose tissue, the functional existence and role of this receptor subtype in man has been controversial. We investigated

Related Content

Why should you have to choose between solvents that are ecological and those that are reliable? Enjoy both at once with our biorenewable and greener solutions. Cyrene™ solvent is a new dipolar aprotic alternative to common REACH restricted solvents, such as N methyl-2-pyrrolidone (NMP) and Dimethylformamide (DMF).

Why should you have to choose between solvents that are ecological and those that are reliable? Enjoy both at once with our biorenewable and greener solutions. Cyrene™ solvent is a new dipolar aprotic alternative to common REACH restricted solvents, such as N methyl-2-pyrrolidone (NMP) and Dimethylformamide (DMF).

Why should you have to choose between solvents that are ecological and those that are reliable? Enjoy both at once with our biorenewable and greener solutions. Cyrene™ solvent is a new dipolar aprotic alternative to common REACH restricted solvents, such as N methyl-2-pyrrolidone (NMP) and Dimethylformamide (DMF).

Why should you have to choose between solvents that are ecological and those that are reliable? Enjoy both at once with our biorenewable and greener solutions. Cyrene™ solvent is a new dipolar aprotic alternative to common REACH restricted solvents, such as N methyl-2-pyrrolidone (NMP) and Dimethylformamide (DMF).

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service