Skip to Content
Merck
All Photos(3)

Key Documents

236705

Sigma-Aldrich

Pentane

Pentane

anhydrous, ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH3
CAS Number:
Molecular Weight:
72.15
Beilstein:
969132
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.21
grade:
anhydrous
Assay:
≥99%
bp:
35-36 °C (lit.)
vapor pressure:
26.76 psi ( 55 °C)
8.28 psi ( 20 °C)
Pricing and availability is not currently available.

grade

anhydrous

Quality Level

vapor density

2.48 (vs air)

vapor pressure

26.76 psi ( 55 °C)
8.28 psi ( 20 °C)

Assay

≥99%

form

liquid

autoignition temp.

500 °F

expl. lim.

~8.3 %

impurities

<0.001% water
<0.005% water (100 mL pkg)

evapn. residue

<0.0005%

refractive index

n20/D 1.358 (lit.)

bp

35-36 °C (lit.)

mp

−130 °C (lit.)

density

0.626 g/mL at 25 °C (lit.)

SMILES string

CCCCC

InChI

1S/C5H12/c1-3-5-4-2/h3-5H2,1-2H3

InChI key

OFBQJSOFQDEBGM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pentane, a linear aliphatic hydrocarbon, is one of the main constituents of light straight-run (LSR) gasoline fraction in crude oils. Its isomerization can improve the octane number of gasoline.[1] This has led to the development of a number of metal catalysts, which can effectively isomerize n-pentane to iso-pentane.[2][3][4]

Application

Pentane may be used as a solvent in the preparation of tetraethynyl silane from tetrakis((trimethylsilyl)ethynyl)silane.[5]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-40.0 °F

Flash Point(C)

-40 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maranda S Cantrell et al.
Foods (Basel, Switzerland), 9(7) (2020-07-10)
Qualitative and semi-quantitative analysis of organosulfides extracted from oil obtained by steam distillation of yellow onions was performed by gas chromatography-mass spectrometry (GC-MS). The extraction efficiency of organosulfides from onion oil was evaluated across four solvents: dichloromethane; diethyl ether; n-pentane;
Re and Pt?Re incorporation in sulfated zirconia as catalysts for n-pentane isomerization
Aboul-Gheit AK, et al
Egyptian Journal of Petroleum, 23(3), 303-314 (2014)
Isomerization of n-pentane over La-Ni-S 2 O 8 2?/ZrO 2-Al 2 O 3 solid superacid catalysts: Deactivation and regeneration.
Song H, et al.
Applied Catalysis A: General, 526, 37-44 (2016)
Supported SO42-/ZrO2 Solid Superacid Catalysts for n-Pentane Isomerization.
Zhao L, et al.
Modern Research in Catalysis, 3(03), 89-89 (2014)
Synthesis and Structure of Tetraethynylsilane and Its Silylated Derivatives.
Geyer FL, et al.
Chemistry?A European Journal , 20(13), 3600-3605 (2014)

Reviews

Active Filters

  1. 1 Ratings-only review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service