Skip to Content
Merck
All Photos(2)

Key Documents

P45605

Sigma-Aldrich

3,3-Dimethyl-2-butanone

97%

Synonym(s):

α,α,α-Trimethylacetone, tert-Butyl methyl ketone, Pinacolone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COC(CH3)3
CAS Number:
Molecular Weight:
100.16
Beilstein:
1209331
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

refractive index

n20/D 1.396 (lit.)

bp

106 °C (lit.)

density

0.801 g/mL at 25 °C (lit.)

SMILES string

CC(=O)C(C)(C)C

InChI

1S/C6H12O/c1-5(7)6(2,3)4/h1-4H3

InChI key

PJGSXYOJTGTZAV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

3,3-Dimethyl-2-butanone is an aliphatic ketone can undergo asymmetric reduction to the corresponding alcohol with diisopinocampheylchloroborane with high enantiomeric excess.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

41.0 °F - closed cup

Flash Point(C)

5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Y Chen et al.
The Journal of organic chemistry, 66(11), 3930-3939 (2001-05-26)
To investigate the effects of electron-donating and electron-withdrawing substituents upon the reaction of porphyrins with osmium tetraoxide, and the pinacol-pinacolone rearrangement of the resulting diols, a series of meso-substituted porphyrins were prepared by total synthesis. Porphyrins with electron-donating substitutents at
Kosuke Namba et al.
Organic letters, 14(5), 1222-1225 (2012-02-22)
An efficient method for the construction of dihydroquinoline derivatives possessing a quaternary carbon center is developed by an application of Hg(OTf)(2)-catalyzed vinylogous semi-pinacol-type rearrangement. The reaction was found to be specifically catalyzed by mercury salt and to proceed via a
Highly efficient asymmetric reduction of. alpha.-tertiary alkyl ketones with diisopinocampheylchloroborane.
Brown H C, et al.
The Journal of Organic Chemistry, 51(17), 3394-3396 (1986)
Richard E Mishler et al.
Chemical communications (Cambridge, England), (41)(41), 6201-6203 (2009-10-15)
We report a new and simple one-pot synthetic method to produce mesoporous silica and nanoporous solid acid catalyst capable of catalyzing pinacole-pinacolone rearrangement and esterification reactions, by preparing a solvent washable phosphonated triblock copolymer template and self-assembling it in the
Direct conversion of arylamines to pinacol boronates: a metal-free borylation process.
Fanyang Mo et al.
Angewandte Chemie (International ed. in English), 49(10), 1846-1849 (2010-02-04)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service