Skip to Content
Merck
All Photos(3)

Key Documents

198099

Sigma-Aldrich

Methyl violet 2B

certified by the Biological Stain Commission

Synonym(s):

Basic Violet 1

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Colour Index Number:
42535
Beilstein:
8464808
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

grade

certified by the Biological Stain Commission

Quality Level

form

crystalline powder

composition

Dye content, 75%

technique(s)

microbe id | staining: suitable

mp

137 °C (dec.) (lit.)

λmax

584 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Cl.C\N=C1/C=C\C(C=C1)=C(/c2ccc(cc2)N(C)C)c3ccc(cc3)N(C)C

InChI

1S/C24H27N3.ClH/c1-25-21-12-6-18(7-13-21)24(19-8-14-22(15-9-19)26(2)3)20-10-16-23(17-11-20)27(4)5;/h6-17H,1-5H3;1H

InChI key

JFTBTTPUYRGXDG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl Violet 2B is a mixture of polymethylated (4,5 and 6) pararosaniline chlorides

Application

Methyl violet 2B has been used to stain human lung fibroblast cells (A549) in in vitro antiviral assay.

Biochem/physiol Actions

Methyl violet 2B is a cationic dye with a triphenylmethane structure. It acts as a selective dye and facilitates the differentiation of Klebsiella pneumoniae from Enterobacter aerogenes and other gram-negative organisms. Methyl violet 2B is harmful if inhaled, swallowed or absorbed through skin, contact may cause damage to skin and eyes. Hence, the dye removal is of great importance.

Suitability

Certified for use in Flemming triple stain with iodine for chromosomes; Newton′s crystal violet-iodine technique for chromatin and nucleoli; Lieb′s amyloid stain.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Annabelle Herrington-Symes et al.
Journal of drug targeting, 25(9-10), 881-890 (2017-08-19)
Increasingly complex proteins can be made by a recombinant chemical approach where proteins that can be made easily can be combined by site-specific chemical conjugation to form multifunctional or more active protein therapeutics. Protein dimers may display increased avidity for
Equilibrium and kinetic studies of methyl violet sorption by agricultural waste
Hameed B H
Journal of Hazardous Materials, 154(1-3), 204-212 (2008)
Methyl Violet: a Selective Agent for Differentiation of
Klebsiella pneumoniae from Enterobacter aerogenes and Other
Gram-Negative Organisms
CAMPBELL L M and ROTH I L
Applied Microbiology, 30, 258-261 (1975)
Expression of soluble and active interferon
consensus in SUMO fusion expression system
in E.coli
Peciak K
Protein Expression and Purification, 99 (2014), 18- 26 (2014)
Removal of cationic dye methyl violet 2B from water by cation exchange membranes
ShiouWu J
Journal of Membrane Science, 309(1-2), 239-245 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service