Skip to Content
Merck
All Photos(2)

Key Documents

195073

Sigma-Aldrich

Mordant Orange 1

Dye content 70 %

Synonym(s):

Alizarin Yellow R, MO1, 5-(4-Nitrophenylazo)salicylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4N=NC6H3-2-(OH)CO2H
CAS Number:
Molecular Weight:
287.23
Colour Index Number:
14030
Beilstein:
962365
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

form

powder

Quality Level

composition

Dye content, 70%

technique(s)

titration: suitable

mp

>300 °C (lit.)

solubility

1 N NH4OH: 1%, clear

λmax

385 nm

ε (extinction coefficient)

≥7000 at 266-276 nm in methanol at 0.006 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

OC(=O)c1cc(ccc1O)N=Nc2ccc(cc2)[N+]([O-])=O

InChI

1S/C13H9N3O5/c17-12-6-3-9(7-11(12)13(18)19)15-14-8-1-4-10(5-2-8)16(20)21/h1-7,17H,(H,18,19)

InChI key

YVJPMMYYRNHJAU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Mordant orange 1 is also known as Alizarin Yellow R, C.I. 14030. Mordant orange 1 is an azo dye. Azo dyes used as biological stains contain an azo group (−N=N−).

Application

Presently, azo dyes are used for coloring many different materials such as textile, leather, and plastics and for manufacturing paints and lacquers.
Presently, azo dyes are used for coloring many different materials such as textile, leather, plastics, food, and pharmaceuticals and for manufacturing paints and lacquers and for printings purposes as well <<<14>>>

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Ming-Chi Tsai et al.
PloS one, 13(12), e0209922-e0209922 (2019-01-01)
Many different time-series methods have been widely used in forecast stock prices for earning a profit. However, there are still some problems in the previous time series models. To overcome the problems, this paper proposes a hybrid time-series model based
Masashi Miyakawa et al.
Scientific reports, 8(1), 12825-12825 (2018-08-29)
Metal oxide TFT fabrication based on a solution-processing method is considered a promising alternative to conventional vacuum processing and has a number of advantages such as low cost, large-area fabrication, and process simplicity. A simple and reliable, direct patterning method
Jihye Kim et al.
Gynecologic oncology, 156(1), 211-221 (2019-11-30)
Cyclin-dependent kinase 7 (CDK7) engages tumor growth by acting as a direct link between the regulation of transcription and the cell cycle. Here, we investigated the clinical significance of CDK7 expression and its potential as a therapeutic target in epithelial
Jianlong Wang et al.
Journal of hazardous materials, 390, 122156-122156 (2020-02-02)
Adsorption technology has been widely applied in water and wastewater treatment, due to its low cost and high efficiency. The adsorption kinetic models have been used to evaluate the performance of the adsorbent and to investigate the adsorption mass transfer
Seth Howes et al.
The Cochrane database of systematic reviews, 4, CD000031-CD000031 (2020-04-23)
Whilst the pharmacological profiles and mechanisms of antidepressants are varied, there are common reasons why they might help people to stop smoking tobacco. Firstly, nicotine withdrawal may produce depressive symptoms and antidepressants may relieve these. Additionally, some antidepressants may have

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service