Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Paraffin Oil
Synonym(s):
Paraffin Oil
CAS Number:
8012-95-1
EC Number:
232-384-2
Products
Product Category
lipids (1)
natural products (1)
solvents (1)
Brand
Sigma-Aldrich (1)
Supelco (2)
Feature
Physical Form
Agency
Quality Segment
Application
Special Grade
Puriss (1)
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
PX0047
Paraffin Oil,
Light
Expand
View Pricing
18512
Paraffin Oil,
puriss., meets analytical specification of Ph. Eur., BP, viscous liquid
Expand
View Pricing
PX0046
Paraffin Oil,
heavy
Expand
View Pricing
76235
Paraffin Oil,
suitable for IR spectroscopy
Expand
View Pricing