Skip to Content
Merck
All Photos(1)

Key Documents

08010

Supelco

Safrole

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

4-Allyl-1,2-methylenedioxybenzene, 5-Allyl-1,3-benzodioxole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10O2
CAS Number:
Molecular Weight:
162.19
Beilstein:
136380
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

vapor pressure

1 mmHg ( 63.8 °C)

product line

TraceCERT®

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

gas chromatography (GC): suitable

refractive index

n20/D 1.537 (lit.)

bp

232-234 °C (lit.)

mp

11.2 °C (lit.)

density

1.095 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

C=CCc1ccc2OCOc2c1

InChI

1S/C10H10O2/c1-2-3-8-4-5-9-10(6-8)12-7-11-9/h2,4-6H,1,3,7H2

InChI key

ZMQAAUBTXCXRIC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Biochem/physiol Actions

Safrole is a naturally-occurring genotoxic compound found in Sassafras root and Areca (betel) quid. It is a hepatocarcinogen, and safrole-DNA adducts have also been seen in oral cancers of Areca users. Metabolites of safrole form adducts with DNA and induce chromasomal aberrations and sister chromatid exchanges.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Principal constituent of sassafras oil

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Muta. 2 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chia-Jung Lee et al.
Molecules (Basel, Switzerland), 25(19) (2020-10-11)
Acne is a common skin condition observed in adolescents. Nutmeg (Myristica fragrans Houtt) (MF) is a well-known traditional Chinese medicine; its major toxic components, safrole and myristicin, are rich in essential oils. Essential oils of MF (MFO) were extracted by
Héctor Carrasco et al.
Molecules (Basel, Switzerland), 17(1), 1002-1024 (2012-01-21)
Twenty one phenylpropanoids (including eugenol and safrole) and synthetic analogues, thirteen of them new compounds, were evaluated for antifungal properties, first with non-targeted assays against a panel of human opportunistic pathogenic fungi. Some structure-activity relationships could be observed, mainly related
Li-Ching Shen et al.
Toxicology letters, 213(3), 309-315 (2012-07-24)
Safrole, a naturally occurring product derived from spices and herbs, has been shown to be associated with the development of hepatocellular carcinoma in rodents. Safrole 2',3'-oxide (SFO), an electrophilic metabolite of safrole, was shown to react with DNA bases to
Chun-Shu Yu et al.
Anticancer research, 32(5), 1671-1679 (2012-05-18)
Safrole, a component of Piper betle inflorescence, is a carcinogen which has been demonstrated to induce apoptosis on human oral cancer HSC-3 cells in vitro and to inhibit HSC-3 cells in xenograft tumor cells in vivo. In our previous study
Le Su et al.
Toxicology letters, 217(2), 129-136 (2012-12-29)
Safrole-2',3'-oxide (SFO) is the major electrophilic metabolite of safrole (4-allyl-1, 2-methylenedioxybenzene), a natural plant constituent found in essential oils of numerous edible herbs and spices and in food containing these herbs, such as pesto sauce, cola beverages and bologna sausages.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service