Skip to Content
Merck
All Photos(1)

Key Documents

09237

Supelco

Myristicin

analytical standard

Synonym(s):

5-Allyl-2,3-(methylendioxy)anisole, 6-Allyl-4-methoxy-1,3-benzodioxole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H12O3
CAS Number:
Molecular Weight:
192.21
Beilstein:
166218
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

COc1cc(CC=C)cc2OCOc12

InChI

1S/C11H12O3/c1-3-4-8-5-9(12-2)11-10(6-8)13-7-14-11/h3,5-6H,1,4,7H2,2H3

InChI key

BNWJOHGLIBDBOB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Myristicin is a major volatile oil and a flavoring plant constituent, reported to be identified in parsley and nutmeg essential oils, black pepper, carrots, etc. It is reported to produce significant psychopharmacological responses.

Application

Myristicin may be used as an analytical reference standard for the quantification of the analyte in the following:
  • Leaf extracts of parsley, dill, and celery grown in microwave fields using high-performance thin-layer chromatography (HPTLC).
  • Rat urine samples using gas chromatography coupled to mass spectrometry (GC-MS).
  • Human serum samples using gas chromatography (GC) with flame ionization detection (FID).

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Repr. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

In vitro and in vivo metabolism of myristicin in the rat
Lee SH, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 705(2), 367-372 (1998)
Simple and rapid determination of myristicin in human serum
Dawidowicz LA and Dybowski PM
Forensic Toxicology, 31(1), 119-123 (2013)
Tahar Smaili et al.
Natural product communications, 6(6), 883-886 (2011-08-06)
The essential oils obtained by hydrodistillation from leaves and fruits of Daucus sahariensis Murb. were analyzed by GC/MS. The main constituents of the essential oil from the leaves were myristicin (34.3%), alpha-pinene (5.4%), cis-chrysanthenyl acetate (5.3%) and epi-alpha-bisabolol (4.8%), and
Nutmeg (Myristica fragrans Houtt.).
Donald G Barceloux
Disease-a-month : DM, 55(6), 373-379 (2009-05-19)
Xin Hang et al.
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica, 32(16), 1669-1675 (2007-11-22)
To analyze the chemical components of the essential oil extracted from the seeds of Myristica fragrans (nutmeg) processed by different methods (steamed with water steam, roasted with flour, sauted with flour, roasted with talcum powder, roasted with loess, and roasted

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service