Skip to Content
Merck
All Photos(1)

Key Documents

06547

Supelco

Methyl formate

analytical standard

Synonym(s):

Formic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCO2CH3
CAS Number:
Molecular Weight:
60.05
Beilstein:
1734623
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

2.1 (vs air)

Assay

≥99.8% (GC)

autoignition temp.

842 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

23 %

refractive index

n20/D 1.343 (lit.)
n20/D 1.344

bp

32-34 °C (lit.)

mp

−100 °C (lit.)

density

0.974 g/mL at 20 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

[H]C(=O)OC

InChI

1S/C2H4O2/c1-4-2-3/h2H,1H3

InChI key

TZIHFWKZFHZASV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl formate is an ester derivative of formic acid.

Application

Methyl formate may be used as a reference standard for the determination of methyl formate in
  • blood and urine by headspace in-tube extraction gas chromatography-mass spectrometry (HS-ITEX-GC−MS)
  • Brazilian sugar-cane spirit by high-resolution gas chromatography (HRGC)
  • pharmaceutical excipients using HS-GC-MS operating under scan mode and selected ion monitoring (SIM) mode

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Flam. Liq. 1 - STOT SE 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-2.2 °F - closed cup

Flash Point(C)

-19 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Formic acid
Ullmann's Encyclopedia of Industrial Chemistry (2000)
Simultaneous determination of formic acid and formaldehyde in pharmaceutical excipients using headspace GC/MS.
del Barrio MA, et al.
Journal of Pharmaceutical and Biomedical Analysis, 41(3), 738-743 (2006)
Identification and dosage by HRGC of minor alcohols and esters in Brazilian sugar-cane spirit.
Boscolo M, et al.
Journal of the Brazilian Chemical Society, 11(1), 86-90 (2000)
Hiroya Yurimoto et al.
Yeast (Chichester, England), 21(4), 341-350 (2004-03-26)
Methyl formate synthesis during growth on methanol by methylotrophic yeasts has been considered to play a role in formaldehyde detoxification. An enzyme that catalyses methyl formate synthesis was purified from methylotrophic yeasts, and was suggested to belong to a family
A Nihlén et al.
International archives of occupational and environmental health, 73(7), 479-487 (2000-11-01)
A toxicokinetic (TK) model was developed to describe the inhalation exposure in humans to methyl formate (MF), a catalyst used in foundries, and to discuss biological monitoring. The TK model consisted of four compartments: MF, the metabolites--methanol (MeOH) and formic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service