Skip to Content
Merck
All Photos(3)

Key Documents

296996

Sigma-Aldrich

Methyl acetate

anhydrous, 99.5%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOCH3
CAS Number:
Molecular Weight:
74.08
Beilstein:
1736662
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.21

grade

anhydrous

Quality Level

vapor density

2.55 (vs air)

vapor pressure

165 mmHg ( 20 °C)

Assay

99.5%

form

liquid

autoignition temp.

936 °F

expl. lim.

16 %

impurities

<0.003% water
<0.005% water (100 mL pkg)

evapn. residue

<0.0003%

refractive index

n20/D 1.361 (lit.)

bp

57-58 °C (lit.)

mp

−98 °C (lit.)

density

0.934 g/mL at 25 °C

SMILES string

COC(C)=O

InChI

1S/C3H6O2/c1-3(4)5-2/h1-2H3

InChI key

KXKVLQRXCPHEJC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl acetate (MA) is an aliphatic ester that can be prepared via carbonylation of dimethyl ether over zeolites. MA is formed as a by-product during the preparation of polyvinyl alcohol from acetic acid and methanol.

Application

Methyl acetate may be used for the preparation of fatty acid methyl esters and triacetin from rapeseed oil via non-catalytic trans-esterification reaction under super-critical conditions.

Other Notes

The article number 296996-6X1L will be discontinued. Please order the single bottle 296996-1L which is physically identical with the same exact specifications.

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

8.6 °F - closed cup

Flash Point(C)

-13 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reaction kinetics and chemical equilibrium of homogeneously and heterogeneously catalyzed acetic acid esterification with methanol and methyl acetate hydrolysis
Popken T, et al.
Industrial & Engineering Chemistry Research, 39(7), 2601-2611 (2000)
Dimethyl ether carbonylation to methyl acetate over HZSM-35.
Liu J, et al.
Catalysis Letters, 139(1-2), 33-37 (2010)
Study on the kinetics of enzymatic interesterification of triglycerides for biodiesel production with methyl acetate as the acyl acceptor.
Xu Y, et al.
Journal of Molecular Catalysis. B, Enzymatic, 32(5), 241-245 (2005)
Long Hu et al.
Nature communications, 12(1), 466-466 (2021-01-22)
All-inorganic CsPbI3 perovskite quantum dots have received substantial research interest for photovoltaic applications because of higher efficiency compared to solar cells using other quantum dots materials and the various exciting properties that perovskites have to offer. These quantum dot devices
Kinetics of transesterification of methyl acetate and n-octanol catalyzed by cation exchange resins.
Liu Y, et al.
Korean Journal of Chemical Engineering, 30(5), 1039-1042 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service