Skip to Content
Merck
All Photos(1)

Key Documents

06084

Supelco

Nitrobenzene

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5NO2
CAS Number:
Molecular Weight:
123.11
Beilstein:
507540
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

4.2 (vs air)

vapor pressure

0.15 mmHg ( 20 °C)
50 mmHg ( 120 °C)

product line

PESTANAL®

Assay

≥99.0% (GC)

autoignition temp.

899 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

40 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.551 (lit.)
n20/D 1.552

bp

210-211 °C (lit.)

mp

5-6 °C (lit.)

density

1.196 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

[O-][N+](=O)c1ccccc1

InChI

1S/C6H5NO2/c8-7(9)6-4-2-1-3-5-6/h1-5H

InChI key

LQNUZADURLCDLV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nitrobenzene is a carcinogenic compound which has been employed extensively in dyes, pesticides and explosives industries.

Application

Nitrobenzene may be used as a reference standard for the analysis of nitrobenzene in soil by gas chromatography (GC) with electron capture detector (ECD) as well as high performance liquid chromatography (HPLC) with ultraviolet (UV) detector and in marine water samples from German Bight by GC coupled to mass spectrometer (MS). It may be used as a reference standard for the determination of nitrobenzene in cigarette smoke by solid phase extraction (SPE) followed by analysis using heart-cutting two-dimensional GC equipped with ECDs.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Carc. 2 - Repr. 1B - STOT RE 1 Inhalation

Target Organs

Blood

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

190.4 °F - closed cup

Flash Point(C)

88 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Determination of seven nitrobenzene compounds in mainstream cigarette smoke with heart-cutting two-dimensional gas chromatography.
Xie F, et al.
Journal of Chromatographic Science, 50(5), 387-392 (2012)
New method for rapid solid-phase extraction of large-volume water samples and its application to non-target screening of North Sea water for organic contaminants by gas chromatography-mass spectrometry.
Weigel S, et al.
Journal of Chromatography A, 912(1), 151-161 (2001)
Yang Mu et al.
Chemosphere, 54(7), 789-794 (2003-11-26)
The reductive degradation of nitrobenzene (NB) by zero-valent iron was investigated. Experimental results showed that the degradation of NB was influenced by pH and NB concentration. The optimum pH value was found to be 3.0 for the reductive degradation of
Determination of nitroaromatic, nitramine, and nitrate ester explosives in soil by gas chromatography and an electron capture detector.
Walsh, Marianne E
Talanta, 54(3), 427-438 (2001)
Iwona Cybulska et al.
Bioresource technology, 118, 30-36 (2012-06-15)
Lignin extracted from prairie cordgrass, switchgrass, and corn stover (using ethyl acetate-ethanol-water organosolv pretreatment) was analyzed and characterized using several methods. These methods included analysis of purity (by determination of Klason lignin, carbohydrate, and ash contents), solubility (with several organic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service