Skip to Content
Merck
All Photos(1)

Key Documents

30-2270

Sigma-Aldrich

Thymol Blue

JIS special grade

Synonym(s):

Thymolsulfonphthalein

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H30O5S
CAS Number:
Molecular Weight:
466.59
Beilstein:
368036
MDL number:
UNSPSC Code:
12171502
PubChem Substance ID:

grade

JIS special grade

form

solid

availability

available only in Japan

visual transition interval

1.2-2.8, red to yellow
8.0-9.2, yellow to blue

mp

221-224 °C (dec.) (lit.)

SMILES string

CC(C)c1cc(c(C)cc1O)C2(OS(=O)(=O)c3ccccc23)c4cc(C(C)C)c(O)cc4C

InChI

1S/C27H30O5S/c1-15(2)19-13-22(17(5)11-24(19)28)27(21-9-7-8-10-26(21)33(30,31)32-27)23-14-20(16(3)4)25(29)12-18(23)6/h7-16,28-29H,1-6H3

InChI key

PRZSXZWFJHEZBJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shangjie Xu et al.
Macromolecular bioscience, 7(8), 968-974 (2007-08-01)
A simple general synthetic concept to build dendritic core-shell architectures with pH-labile linkers based on hyperbranched PEI cores and biocompatible PEG shells is presented. Using these dendritic core-shell architectures as nanocarriers, the encapsulation and transport of polar dyes of different
J G Eley et al.
The Journal of pharmacy and pharmacology, 41(12), 858-860 (1989-12-01)
The interaction of low density lipoprotein (LDL) with two model dyes in the presence or absence of non-ionic surfactants has been studied. The water soluble dye, thymol blue, only weakly interacts with LDL to cause slight increases in particle size
P Balderas-Hernández et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 66(1), 68-73 (2006-07-11)
This paper presents the results concerning the determination of the formation constants of the complexes between thymol blue, TB, and the metal ions Pb(II) or Cd(II). The experimental procedure was carried out in the presence of a nitrogen atmosphere at
N Gayathri et al.
Journal of AOAC International, 84(4), 1065-1069 (2001-08-15)
A simple and sensitive spectrophotometric method was developed for the determination of trace amounts of sulfur dioxide. The method is based on the reaction of SO2 with a known excess of ICI as the oxidant. The unreacted ICI iodinates thymol
Yong-Doo Park et al.
Journal of protein chemistry, 22(5), 473-480 (2003-12-24)
Sensitive assay methods for tyrosinase are essential not only for the understanding the process of pigment production but also for the development of effective inhibitors of tyrosinase. To develop an efficient assay method, we applied thymol blue to reaction mixtures.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service