Skip to Content
Merck
All Photos(1)

Key Documents

19-4497

Sigma-Aldrich

Metronidazole

SAJ first grade, ≥99.0%

Synonym(s):

2-Methyl-5-nitroimidazole-1-ethanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H9N3O3
CAS Number:
Molecular Weight:
171.15
Beilstein:
611683
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
grade:
SAJ first grade

grade

SAJ first grade

Assay

≥99.0%

form

solid

availability

available only in Japan

dilution

(for analytical testing)

mp

159-161 °C (lit.)

antibiotic activity spectrum

Gram-negative bacteria
parasites

Mode of action

DNA synthesis | interferes

SMILES string

CC1=NC=C([N+]([O-])=O)N1CCO

InChI

1S/C6H9N3O3/c1-5-7-4-6(9(11)12)8(5)2-3-10/h4,10H,2-3H2,1H3

InChI key

VAOCPAMSLUNLGC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: imidazole

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Muta. 1B - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T H I Brummer et al.
BJOG : an international journal of obstetrics and gynaecology, 120(10), 1269-1276 (2013-06-22)
To evaluate cefuroxime and metronidazole antibiotic prophylaxis. Observational nonrandomised 1-year prospective cohort study. Fifty-three hospitals in Finland. A total of 5279 women undergoing hysterectomy for benign indications, with cefuroxime given to 4301 and metronidazole given to 2855. Excluding other antibiotics
Christina M Surawicz et al.
The American journal of gastroenterology, 108(4), 478-498 (2013-02-27)
Clostridium difficile infection (CDI) is a leading cause of hospital-associated gastrointestinal illness and places a high burden on our health-care system. Patients with CDI typically have extended lengths-of-stay in hospitals, and CDI is a frequent cause of large hospital outbreaks
Sonja Löfmark et al.
Clinical infectious diseases : an official publication of the Infectious Diseases Society of America, 50 Suppl 1, S16-S23 (2010-01-14)
Metronidazole has been used for the treatment of infections for >45 years and is still successfully used for the treatment of trichomoniasis, amoebiasis, and giardiasis. Anaerobic bacterial infections caused by Bacteroides species, fusobacteria, and clostridia respond favorably to metronidazole therapy.
Konstantinos Z Vardakas et al.
International journal of antimicrobial agents, 40(1), 1-8 (2012-03-09)
The objective of this review was to evaluate the frequency of treatment failure and recurrence of Clostridium difficile infection (CDI) following treatment with vancomycin or metronidazole in recently performed studies (last 10 years). Searches in PubMed and Scopus were performed
Dina Zandbergen et al.
Journal of periodontology, 84(3), 332-351 (2012-05-23)
The treatment of periodontitis frequently begins with a non-surgical phase that includes scaling and root planing (SRP) and, on occasion, the use of systemic antibiotics. The goal of this review is to systematically evaluate the data concerning the effect of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service