Merck
  • The clinical effect of scaling and root planing and the concomitant administration of systemic amoxicillin and metronidazole: a systematic review.
The clinical effect of scaling and root planing and the concomitant administration of systemic amoxicillin and metronidazole: a systematic review.
Journal of periodontology (2012-05-23)
Dina Zandbergen, Dagmar E Slot, Charles M Cobb, Fridus A Van der Weijden
ABSTRACT
The treatment of periodontitis frequently begins with a non-surgical phase that includes scaling and root planing (SRP) and, on occasion, the use of systemic antibiotics. The goal of this review is to systematically evaluate the data concerning the effect of the concomitant administration of amoxicillin and metronidazole adjunctive to SRP in adults who are otherwise healthy. The PubMed-MEDLINE, Cochrane-Central, and EMBASE databases were searched to April 1, 2012, to identify appropriate studies. Probing depth (PD), clinical attachment level (CAL), bleeding on probing, and plaque index were selected as outcome variables. Based on the extracted mean values and number of individuals, changes in weighted means were calculated and a meta-analysis conducted. The search yielded 526 unique titles and abstracts. Ultimately, 35 studies were selected, describing 28 clinical trials meeting the eligibility criteria. The full-mouth weighted mean change for PD showed an improvement of 1.41 mm. The full-mouth weighted mean change for CAL showed a gain of 0.94 mm. Systemic antimicrobial therapy using a combination of amoxicillin and metronidazole as an adjunct to SRP can enhance the clinical benefits of non-surgical periodontal therapy in adults who are otherwise healthy.
Product Description
Supelco
Metronidazole, VETRANAL®, analytical standard
Sigma-Aldrich
Metronidazole, BioXtra
Supelco
Metronidazole, analytical standard
Sigma-Aldrich
Metronidazole, SAJ first grade, ≥99.0%
Supelco
Metronidazole, Pharmaceutical Secondary Standard; Certified Reference Material
Metronidazole, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Amoxicillin, 95.0-102.0% anhydrous basis