Skip to Content
Merck
All Photos(1)

Key Documents

09-1680

Sigma-Aldrich

2-Ethoxyethanol

SAJ special grade, ≥99.0%

Synonym(s):

Ethyl glycol, Ethylene glycol monoethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OCH2CH2OH
CAS Number:
Molecular Weight:
90.12
Beilstein:
1098271
EC Number:
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:

grade

SAJ special grade

vapor density

3.1 (vs air)

vapor pressure

3.8 mmHg ( 20 °C)

Assay

≥99.0%

form

liquid

autoignition temp.

460 °F

expl. lim.

14 %

availability

available only in Japan

refractive index

n20/D 1.407 (lit.)

bp

135 °C (lit.)

mp

−90 °C (lit.)

density

0.93 g/mL at 25 °C (lit.)

SMILES string

CCOCCO

InChI

1S/C4H10O2/c1-2-6-4-3-5/h5H,2-4H2,1H3

InChI key

ZNQVEEAIQZEUHB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 3 - Repr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

104.0 °F - closed cup

Flash Point(C)

40 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I A Adedara et al.
Andrologia, 45(2), 111-119 (2012-06-20)
Endocrine disrupting chemicals cause reproductive dysfunction by interacting with intricate regulation and cellular processes involve in spermatogenesis. This study investigated the probable mechanism of action of ethylene glycol monoethyl ether (EGEE) as an antiandrogenic compound as well as the effects
Gargi Bagchi et al.
International journal of andrology, 31(2), 269-274 (2008-01-09)
Methoxyacetic acid (MAA), the active biological oxidation product of the industrial solvent ethylene glycol monomethyl ether (EGME), causes acute toxicity in several species including humans. MAA primarily affects tissues with rapidly dividing cells and high rates of energy metabolism, including
Chang Yong Yoon et al.
The Journal of veterinary medical science, 65(2), 207-212 (2003-03-26)
The effects of ethylene glycol monoethyl ether (EGEE) on testicular cell populations in rats were investigated by a flow cytometric method. Rats were administered by gavage with EGEE at the various doses of 0 (saline alone), 100, 200, 400, and
David J Lockley et al.
Toxicology and applied pharmacology, 180(2), 74-82 (2002-04-24)
Percutaneous absorption and cutaneous metabolism of 2-ethoxyethanol were assessed in vivo and with an in vitro flow-through diffusion system. Topical application of undiluted (14)C-ethoxyethanol to occluded rat skin in vivo resulted in 25% of the dose being absorbed after 24
Randa A El-Zein et al.
Archives of environmental health, 57(4), 371-376 (2003-01-18)
Glycol ethers are known reproductive and developmental toxins in laboratory animals, but little is known about their genotoxic effects in humans. In the current article, the authors tested the hypothesis that human in utero exposure to ethylene glycol monomethyl ether

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service