Merck
  • Adsorption of Cu(II), Hg(II), and Ni(II) ions by modified natural wool chelating fibers.
Adsorption of Cu(II), Hg(II), and Ni(II) ions by modified natural wool chelating fibers.
Journal of hazardous materials (2009-12-08)
M Monier, D M Ayad, A A Sarhan
ABSTRACT
The graft copolymerization of ethyl acrylate (EA) onto natural wool fibers initiated by potassium persulphate and Mohr's salt redox initiator system in limited aqueous medium was carried out in heterogeneous media. Ester groups of the grafted copolymers were partially converted into hydrazide function groups followed by hydrazone formation through reaction with isatin. Also the application of the modified fibers for metal ion uptake was studied using Cu(II), Hg(II) and Ni(II). The modified chelating fibers were characterized using FTIR spectroscopy, SEM and X-ray diffraction.
Sigma-Aldrich
Ethyl acrylate, contains 10-20 ppm MEHQ as inhibitor, 99%
Sigma-Aldrich
Ethyl acrylate, SAJ first grade, ≥99.0%
Supelco
Ethyl acrylate, analytical standard
Sigma-Aldrich
Ethyl acrylate, ≥99.5%, stabilized