Skip to Content
Merck
All Photos(1)

Documents

09-0410

Sigma-Aldrich

Eriochrome® Black T solution

Synonym(s):

Mordant Black 11

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H12N3NaO7S
CAS Number:
Molecular Weight:
461.38
Beilstein:
4121162
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:

availability

available only in Japan

SMILES string

[Na+].Oc1cc(c2cc(ccc2c1\N=N\c3ccc4ccccc4c3O)[N+]([O-])=O)S([O-])(=O)=O

InChI

1S/C20H13N3O7S.Na/c24-17-10-18(31(28,29)30)15-9-12(23(26)27)6-7-14(15)19(17)22-21-16-8-5-11-3-1-2-4-13(11)20(16)25;/h1-10,24-25H,(H,28,29,30);/q;+1/p-1/b22-21+;

InChI key

AMMWFYKTZVIRFN-QUABFQRHSA-M

Looking for similar products? Visit Product Comparison Guide

Suitability

for titration

Legal Information

Eriochrome is a registered trademark of Huntsman Petrochemical, LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hong Yao et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 23(6), 677-682 (2007-06-19)
An electropolymerized film of eriochrome black T (EBT) has been prepared at a glassy carbon electrode (GCE) by cyclic voltammetry (CV). The poly(EBT) membrane at GCE exhibits an excellent electrocatalytic activity towards the oxidation of epinephrine (EP), ascorbic acid (AA)
Liqing Lin et al.
Talanta, 80(5), 2113-2119 (2010-02-16)
A novel electrochemical biosensor is described for detection of breakpoint cluster region gene and a cellular abl (BCR/ABL) fusion gene in chronic myelogenous leukemia (CML) by using thiolated-hairpin locked nucleic acids (LNA) as the capture probe. The hairpin LNA probe
Li-Tai Jin et al.
Proteomics, 6(8), 2334-2337 (2006-02-24)
A quick, sensitive, and MALDI-TOF MS compatible silver staining method, namely Eriochrome black T (EBT)-silver method, is described. The method can detect 0.05-0.2 ng protein within 60 min in SDS-PAGE gels. EBT dye was used as a silver ion sensitizer
R T Gettar et al.
Journal of chromatography. A, 855(1), 111-119 (1999-10-09)
The use of Eriochrome Black T in an alkaline, 40% methanol solution was found to be appropriate as post-column reagent for the determination of rare earths by ion chromatography. Detection of individual lanthanides and lanthanum was carried out at 512
T Kvernmo et al.
The international journal of biochemistry & cell biology, 28(3), 303-309 (1996-03-01)
Due to the central role of sorbitol dehydrogenase in diabetic cataract, it is important to examine this enzyme's interaction with different inhibitory compounds such as dyes. The aim of the study was to investigate the binding of Cibacron Blue and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service