Skip to Content
Merck
All Photos(1)

Documents

09-0400

Sigma-Aldrich

Eriochrome® Black T

SAJ special grade

Synonym(s):

Mordant Black 11

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H12N3NaO7S
CAS Number:
Molecular Weight:
461.38
Colour Index Number:
14645
Beilstein:
4121162
EC Number:
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:

grade

SAJ special grade

availability

available only in Japan

SMILES string

[Na+].Oc1cc(c2cc(ccc2c1\N=N\c3ccc4ccccc4c3O)[N+]([O-])=O)S([O-])(=O)=O

InChI

1S/C20H13N3O7S.Na/c24-17-10-18(31(28,29)30)15-9-12(23(26)27)6-7-14(15)19(17)22-21-16-8-5-11-3-1-2-4-13(11)20(16)25;/h1-10,24-25H,(H,28,29,30);/q;+1/p-1/b22-21+;

InChI key

AMMWFYKTZVIRFN-QUABFQRHSA-M

Looking for similar products? Visit Product Comparison Guide

Legal Information

Eriochrome is a registered trademark of Huntsman Petrochemical, LLC

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hong Yao et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 23(6), 677-682 (2007-06-19)
An electropolymerized film of eriochrome black T (EBT) has been prepared at a glassy carbon electrode (GCE) by cyclic voltammetry (CV). The poly(EBT) membrane at GCE exhibits an excellent electrocatalytic activity towards the oxidation of epinephrine (EP), ascorbic acid (AA)
Liqing Lin et al.
Talanta, 80(5), 2113-2119 (2010-02-16)
A novel electrochemical biosensor is described for detection of breakpoint cluster region gene and a cellular abl (BCR/ABL) fusion gene in chronic myelogenous leukemia (CML) by using thiolated-hairpin locked nucleic acids (LNA) as the capture probe. The hairpin LNA probe
J L Seltzer et al.
Collagen and related research, 7(6), 399-407 (1987-12-01)
The intracellular degradation of interstitial collagen is accomplished by two neutral metalloproteases, collagenase and gelatinase. Both enzymes are inhibited by metal chelating agents, by certain sulfhydryl reagents, and by similar protein inhibitors. Here, we demonstrate that the dye eriochrome black
Detection of proteins in polyacrylamide gels using eriochrome black T and rhodamine B.
D W Jung et al.
Analytical biochemistry, 263(1), 118-120 (1998-09-29)
R T Gettar et al.
Journal of chromatography. A, 855(1), 111-119 (1999-10-09)
The use of Eriochrome Black T in an alkaline, 40% methanol solution was found to be appropriate as post-column reagent for the determination of rare earths by ion chromatography. Detection of individual lanthanides and lanthanum was carried out at 512

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service